Skip to Content

1.11169

Nitrate Test Kit, colorimetric

10-150 mg/L (NO₃⁻), MQuant®

Synonym(s):

Nitrate Visual Test Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nitrate Test in freshwater, colorimetric, 10-150 mg/L (NO3-), for use with MCOLORTEST®

specific analyte(s)

nitrate

Quality Segment

measuring range

10-150 mg/L (NO3-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card 10 - 25 - 50 - 75 - 100 - 125 - 150 mg/l NO MQuant®
This MQuant® Nitrate test (formally MColortest®) is suitable for the colorimetric determination using the easy to use color card. The reagents are added to the sample, resulting in a color reaction. The concentration is determined by allocating the color to a value on a reference scale.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Nitrate Test in freshwater


signalword

Danger

wgk

WGK 3

target_organs

Kidney,Lungs,Bone

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Pyr. Sol. 1 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Inhalation

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library