Skip to Content
Merck
All Photos(2)

Key Documents

1.10013

Supelco

Sulfite Test Strips, colorimetric

greener alternative

10-400 mg/L (SO₃²⁻), MQuant®, for use with MQuant® StripScan App

Synonym(s):

Sulfite chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Sulfite Test, colorimetric, 10-400 mg/L (SO32-), for use with MQuant® StripScan App, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

sulfite

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

measuring range

10-400 mg/L (SO32-)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

greener alternative category

storage temp.

2-8°C

General description

Method: Colorimetric, with test strips, 10 - 40 - 80 - 180 - 400 mg/L (SO32-), MQuant®

The MQuant® Sulfite test strips are suitable for food analysis, particularly in the testing of wine and fruit juices, sulfurated foods (e.g. fresh and dried fruits), fish, shellfish, crustaceans and minced meat products. Sulfite ions react with a mixture of potassium hexacyanoferrate(II), zinc sulfate, and sodium nitroprusside to form a red compound. The sulfite concentration is measured semi-quantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the sulfite concentration and a possible dilution factor prior to using a quantitative method such as a Reflectoquant® or Spectroquant® Sulfite test.

Design for sustainability benefits:

We continue to demonstrate our commitment to our planet and a sustainable future by exploring ways to improve existing products in our portfolio. We have recently reviewed the current line of MQuant® test strips to identify key opportunities to minimize impact without compromising on the product performance.

This product has benefited from:

  • Test strip with reduction
  • Recycled aluminum tube
  • Digital instruction leaflet

Learn more about the initiative: Small Changes to Test Strips for Big Impact.

Linkage

Replaces: 10013-1; 10013

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Preparation of a standard solution for Sulfite

Related Content

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service