1.09556
pH-indicator paper pH 5.4 - 7.0 Special indicator
pH range 5.4-7.0, roll L 4.8 m, with color scale, graduations and accuracy: 0.4 pH unit, MQuant®
Synonym(s):
pH chemical test paper paper pH 5.4 - 7.0 Special indicator, pH test paper paper pH 5.4 - 7.0 Special indicator
About This Item
product name
pH-indicator paper pH 5.4 - 7.0 Special indicator, roll L 4.8 m , with color scale, pH range 5.4-7.0, graduations and accuracy accuracy: 0.4 pH unit, MQuant®
product line
MQuant®
Quality Level
feature
non-bleeding: no
graduations and accuracy measuring range
accuracy: 0.4 pH unit
roll L
4.8 m , with color scale
pH range
5.4-7.0
detection method
colorimetric
storage temp.
15-25°C
General description
MQuant® pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences, such as moisture, light, and ambient gases. These pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. The paper is always ready to use; it is easily pulled out of the plastic reel and can be torn off in the necessary length. The pH value is measured semi-quantitatively by visual comparison of the color formed on the paper with the fields of the color scale.
Analysis Note
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service