Skip to Content
Merck
All Photos(1)

Key Documents

1.08917

Supelco

Zirconium(IV) oxide chloride octahydrate

for analysis EMSURE®

Synonym(s):

Zirconium(IV) oxide chloride octahydrate, Zirconyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2OZr · 8H2O
CAS Number:
Molecular Weight:
322.25
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-717-9
NACRES:
NA.21
Assay:
≥97.0% (gravimetric)
form:
solid
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥97.0% (gravimetric)

form

solid

potency

3500 mg/kg LD50, oral (Rat)

pH

1 (20 °C, 50 g/L in H2O)

density

1.91 g/cm3 at 20 °C

bulk density

800 kg/m3

anion traces

sulfate (SO42-): ≤0.015%

cation traces

Fe: ≤0.001%
Ti: ≤0.005%
heavy metals (as Pb): ≤0.001%

storage temp.

15-25°C

SMILES string

[Zr]=O.Cl.Cl.O.O.O.O.O.O.O.O

InChI

1S/2ClH.8H2O.O.Zr/h2*1H;8*1H2;;

InChI key

FUBACIUATZGHAC-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (gravimetrically): ≥ 97.0 %
Sulfate (SO₄): ≤ 0.015 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.001 %
Ti (Titanium): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service