Skip to Content
Merck

Skip To

1.07656

Lactose monohydrate

powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF

Synonym(s):

Lactose monohydrate, Milk sugar, Lactobiose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
Molecular Weight:
360.32
MDL number:
UNSPSC Code:
12352201
EC Index Number:
200-559-2
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, ChP, JP, NF, Ph. Eur.

product line

EMPROVE® ESSENTIAL

form

fine powder, powder

autoignition temp.

390 °C

potency

>10000 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Olefin Metathesis

pH

4.0-6.5 (20 °C, 100 g/L in H2O)

mp

202 °C

solubility

161 g/L

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

15-25°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@H]([C@H]1CO)O)O)O)O[C@@H]2[C@H](O[C@@H]([C@@H]([C@H]2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5+,6+,7-,8-,9-,10-,11+,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-XLOQQCSPSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Lactose is a commonly used ingredient in food and pharmaceutics. In pharmaceutical applications, lactose has been used primarily for solid oral dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1081951076601356701
description

powder, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF

description

(milk sugar) special grade for tabletting, EMPROVE® ESSENTIAL, Ph. Eur., BP, ChP, JP, NF

description

for biochemistry

description

United States Pharmacopeia (USP) Reference Standard

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

application(s)

-

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

agency

BP, JP, Ph. Eur., ChP, NF

agency

BP, ChP, JP, NF, Ph. Eur.

agency

-

agency

USP

form

fine powder

form

fine powder

form

powder

form

-

solubility

161 g/L

solubility

-

solubility

161 g/L

solubility

-

product line

EMPROVE® ESSENTIAL

product line

EMPROVE® ESSENTIAL

product line

-

product line

-

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

-


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters