Skip to Content
Merck
All Photos(2)

Key Documents

1.06579

Supelco

di-Sodium hydrogen phosphate dodecahydrate

for analysis EMSURE® ISO,Reag. Ph Eur

Synonym(s):

Sodium phosphate dibasic dodecahydrate, Sodium monohydrogen phosphate, Sodium phosphate dibasic, sec-Sodium phosphate, Disodium hydrogen phosphate dodecahydrate, Disodium phosphate, di-Sodium hydrogen phosphate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4 · 12H2O
CAS Number:
Molecular Weight:
358.14
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-448-7
NACRES:
NA.21
Assay:
99.0-102.0% (alkalimetric)
form:
solid
solubility:
218 g/L

Agency

reag. ISO
reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

99.0-102.0% (alkalimetric)

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

composition

Water, 57-61%

impurities

≤2.5% Sodium Dihydrogene phospate

pH

9.0-9.3 (20 °C, 50 g/L in H2O)

solubility

218 g/L

density

1.52 g/cm3 at 20 °C

bulk density

800‑900 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

As: ≤0.00005%
Cu: ≤0.0002%
Fe: ≤0.0005%
K: ≤0.005%
N: ≤0.001%
Pb: ≤0.0005%
heavy metals (as Pb): ≤0.0005%

storage temp.

15-25°C

SMILES string

O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P.12H2O/c;;1-5(2,3)4;;;;;;;;;;;;/h;;(H3,1,2,3,4);12*1H2/q2*+1;;;;;;;;;;;;;/p-2

InChI key

DGLRDKLJZLEJCY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Linkage

Replaces: SX0718; SX0718-1

Analysis Note

Assay (alkalimetric): 99.0 - 102.0 %
Identity: passes test
Appearance of solution: passes test
Sodium Dihydrogene phospate: ≤ 2.5 %
pH-value (5 %; water): 9.0 - 9.3
Chloride (Cl): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
As (Arsenic): ≤ 0.00005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Pb (Lead): ≤ 0.0005 %
Reducing substances: passes test
Water: 57.0 - 61.0 %
Corresponds to ISO,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

LC-MS/MS-based determination of 19 sulfonamides, 13 quinolones, and 3 tetracycline drug residues in pork meat samples using Supel™ Swift HLB for extraction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service