Skip to Content
Merck
All Photos(1)

Key Documents

1.06363

Supelco

Sodium bromide

99.995 Suprapur®

Synonym(s):

Sodium bromide, Bromo sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BrNa
CAS Number:
Molecular Weight:
102.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-599-9
NACRES:
NA.21
Assay:
99.995%
grade:
for inorganic trace analysis
solubility:
946 g/L
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

vapor pressure

1.3 hPa ( 806 °C)

Assay

99.995%

potency

3500 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

5.74 (22.5 °C, 430 g/L in H2O)

solubility

946 g/L

density

3.2 g/cm3 at 25 °C

bulk density

1750 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Br-]

InChI

1S/BrH.Na/h1H;/q;+1/p-1

InChI key

JHJLBTNAGRQEKS-UHFFFAOYSA-M

Related Categories

Analysis Note

Purity (metallic): ≥ 99.995 %
Assay (argentometric): ≥ 99.0 %
Phosphate (PO₄) (as P): ≤ 10 ppm
Sulfate (SO₄) (as S): ≤ 20 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 5.0 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cs (Cesium): ≤ 5 ppm
Cu (Copper): ≤ 0.050 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 10 ppm
La (Lanthanum): ≤ 0.010 ppm
Li (Lithium): ≤ 0.1 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.050 ppm
Ni (Nickel): ≤ 0.05 ppm
Pb (Lead): ≤ 0.05 ppm
Rb (Rubidium): ≤ 5 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.05 ppm
Tl (Thallium): ≤ 0.05 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.05 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service