1.06223
1-Naphthol
GR for analysis
Synonym(s):
1-Naphthol, 1-Hydroxynaphthalene
About This Item
Recommended Products
vapor pressure
1.3 hPa ( 94 °C)
Quality Level
Assay
≥99% (GC)
form
solid
autoignition temp.
510 °C
potency
1870 mg/kg LD50, oral (Rat)
880 mg/kg LD50, skin (Rabbit)
bp
288 °C/1013 hPa
mp
94-96 °C
transition temp
flash point 125 °C
solubility
0.1 g/L
density
1.28 g/cm3 at 20 °C
bulk density
450 kg/m3
storage temp.
2-30°C
SMILES string
Oc1c2c(ccc1)cccc2
InChI
1S/C10H8O/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7,11H
InChI key
KJCVRFUGPWSIIH-UHFFFAOYSA-N
Analysis Note
2-Naphthol (GC, area%): ≤ 0.20 % (a/a)
Naphthalene (GC, area%): ≤ 0.20 % (a/a)
Melting range (lower value): ≥ 95 °C
Melting range (upper value): ≤ 97 °C
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test
Chloride (Cl): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Iron (Fe): ≤ 0.001 %
Sulfated ash: ≤ 0.05 %
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1A - STOT SE 2 Oral - STOT SE 3
Target Organs
Kidney, Respiratory system
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
257.0 °F - closed cup
Flash Point(C)
125 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service