Skip to Content
Merck

Skip To

1.04672

Synthechol® Synthetic Cholesterol

EMPROVE® EXPERT, Ph. Eur., BP, JP, NF

Synonym(s):

Cholesterol, 3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
UNSPSC Code:
12352104
Beilstein/REAXYS Number:
1915888
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

agency

BP, JP, NF, Ph. Eur.

product line

EMPROVE® EXPERT

form

powder

bp

360 °C (lit.)

mp

~141 °C, 147-149 °C (lit.)

density

1.067 g/mL at 25 °C (lit.)

suitability

suitable for (Suitable for formulation and manufacturing use)

application(s)

advanced drug delivery
controlled release
liquid formulation
parenterals
pharmaceutical

storage temp.

15-25°C

SMILES string

CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

General description

A GMP-scalable process which yields a high-quality lipid is essential to lipidic drug product development and manufacturing. We are highly specialized in synthesizing top-quality GMP lipids for pharmaceutical applications and gene therapy, focusing on tailored manufacturing to meet your specific needs. We also offer an off-the-shelf catalog with optimized stability, solubility, and handling characteristics.
Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Cholesterol is a major component of all biological cell membranes and is used as a stabilizer of lipid bilayers in many lipid-based formulations for the delivery of different APIs, such as small molecules and nucleic acids.
Synthetic cholesterol, Ph Eur, BP, JP, NF is ultra-pure and therefore suitable for divers formulations applications. Due to low batch to batch variability, very high purity, and lack of biological components, this product is optimal for highly sensitive applications such as nucleic acid delivery.
With a newly optimized manufacturing process, this product is available in quantities from grams to tons to support your development progress. A dual sourcing option is also possible.

Biochem/physiol Actions

Major component of all biological membranes; ~25% of total brain lipid is cholesterol.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany
SyntheChol is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The substance 1.04672 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.04675C2155000700100P
biological source

synthetic

biological source

synthetic

biological source

-

biological source

-

storage temp.

15-25°C

storage temp.

no temp limit

storage temp.

2-8°C

storage temp.

−20°C

suitability

suitable for (Suitable for formulation and manufacturing use)

suitability

-

suitability

-

suitability

-

form

powder

form

-

form

-

form

powder

Quality Level

500

Quality Level

100

Quality Level

-

Quality Level

-

bp

360 °C (lit.)

bp

360 °C (lit.)

bp

360 °C (lit.)

bp

-


Still not finding the right product?

Explore all of our products under Synthechol® Synthetic Cholesterol


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters