Skip to Content

1.04161

Ammonium sulfate

granulated, EMPROVE® EXPERT, ACS, ChP, NF

Synonym(s):

Mascagnite

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
UNSPSC Code:
12352314
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

GMO free, Halal, Kosher

agency

ChP, NF

product line

EMPROVE® EXPERT

form

solid

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

application(s)

liquid formulation
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

2-30°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

General description

Standard bulk materials often show a tendency for caking, clumping and dust formation which results in laborious weighing and manual de-caking processes with a negative impact on manufacturing efficiency. Due to its granulated form, Ammonium Sulfate granulated is free-flowing and easy to handle. Its greatly reduced caking behavior facilitates processability, speeds up manufacturing processes and increases operator safety.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ammonium sulfate is widely used as a precipitant in biopharma manufacturing and as a excipient in liposomal formulations.
Due to its low microbial and endotoxin limits, Ammonium sulfate Emprove® Expert is especially suitable for high-risk applications.

Packaging

1.04161.0001 / 3x 1 kg / PE bottle
1.04161.1000 / 1 kg / PE bottle
1.04161.9026 / 25 kg / PE bag in PE drum

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Ammonium sulfate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.