Skip to Content
Merck
All Photos(1)

Key Documents

1.03967

SAFC

Iron(II) sulfate hydrate

(about 87% FeSO₄) extra pure Ph.Eur.,BP,USP

Pharma Manufacturing

Synonym(s):

Iron(II) sulfate hydrate, Ferrosulfate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeO4S · xH2O
CAS Number:
Molecular Weight:
151.91 (anhydrous basis)
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-753-5
Pricing and availability is not currently available.

Agency

BP
Ph. Eur.
USP

Quality Level

form

solid

potency

319 mg/kg LD50, oral (Rat)

pH

2.5-3.8 (20 °C, 50 g/L in H2O)

mp

300 °C (Elimination of water of crystallisation)

density

2.97 g/cm3

bulk density

850 kg/m3

application(s)

pharma/biopharma processes

storage temp.

15-25°C

SMILES string

[Fe+2].[S](=O)(=O)([O-])[O-].O

InChI

1S/Fe.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

XBDUTCVQJHJTQZ-UHFFFAOYSA-L

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Iron (II) sulfate hydrate is used as a precursor to iron compounds and as a reducing agent.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service