Skip to Content
Merck
All Photos(1)

Key Documents

1.03132

Supelco

1,4-Dioxane

for liquid chromatography LiChrosolv®

Synonym(s):

1,4-Dioxane, Glycolethylether, 1,4-Diethylene dioxide, 1,4-Dioxacyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
MDL number:
UNSPSC Code:
12191504
EC Index Number:
204-661-8
NACRES:
NA.03
technique(s):
HPLC: suitable
bp:
101.5 °C/1013 hPa
vapor pressure:
41 hPa ( 20 °C)

vapor pressure

41 hPa ( 20 °C)

Quality Level

description

isocratic

product line

LiChrosolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

300 °C

potency

5200 mg/kg LD50, oral (Rat)
7600 mg/kg LD50, skin (Rabbit)

expl. lim.

1.7-25.2 % (v/v)

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.02% Water

evapn. residue

≤2.0 mg/L

transmittance

245 nm, ≥50%
270 nm, ≥80%
300 nm, ≥98%

pH

6-8 (20 °C, 500 g/L in H2O)

bp

101.5 °C/1013 hPa

mp

12 °C

transition temp

flash point 11 °C

density

1.03 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1CCOCC1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • Cross-Polarization Solid-State NMR Quantification of Species within Pores of Metal-Organic Frameworks: Though not directly linked to 1,4-dioxane, this study employs methodologies that could be applicable for the analysis of dioxane and other similar molecules within complex structures, relevant for chemists and material scientists engaged in molecular characterization (Wu & Xu, 2024).

  • Photosensitized covalent organic framework as a light-induced oxidase mimic for colorimetric detection of uric acid: This innovative approach using a framework that could potentially be adapted for 1,4-dioxane detection underscores the intersection of material science and analytical chemistry, which could intrigue researchers developing new analytical methods (Wang et al., 2024).

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.8 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.02 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 245 nm): ≥ 50 %
Transmission (at 270 nm): ≥ 80 %
Transmission (from 300 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service