Skip to Content
Merck

1.02413

L-Tyrosine disodium salt dihydrate

suitable for cell culture

Synonym(s):

L-Tyrosine disodium salt dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H9NNa2O3 · 2H2O
CAS Number:
Molecular Weight:
261.18
MDL number:
UNSPSC Code:
41106213
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


technique(s)

cell culture | mammalian: suitable

Quality Level

storage temp.

2-8°C

SMILES string

[Na+].[Na+].N[C@@H](Cc1ccc(cc1)[O-])C(=O)[O-].O.O

InChI

1S/C9H11NO3.2Na.2H2O/c10-8(9(12)13)5-6-1-3-7(11)4-2-6;;;;/h1-4,8,11H,5,10H2,(H,12,13);;;2*1H2/q;2*+1;;/p-2/t8-;;;;/m0..../s1

InChI key

WFTCFVUQORELJZ-USHJOAKVSA-L

General description

for cell culture media (not for dietary use)


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
102413100004027269846015
102413010004054839017278