Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74
EC Index Number:
265-363-1
Product Name
Casein hydrolysate, acid hydrolyzed, suitable for microbiology
biological source
bovine milk
Quality Segment
assay
7.0-8.5% amino-nitrogen basis (purity)
form
powder
shelf life
limited shelf life, expiry date on the label
packaging
pkg of 5 kg, pkg of 500 g
storage condition
(Tightly closed)
pH
4.7-5.7 (20 °C, 50 g/L in H2O)
solubility
310 g/L
bulk density
600 kg/m3
application(s)
microbiology
storage temp.
15-25°C
General description
Casein hydrolysate is commonly used in microbiology as a nitrogen source and a supplement in microbial culture media. It is produced by the digestion of casein with hydrochloric acid and contains a complex mixture of peptides and amino acids. Casein hydrolysate is particularly useful in media formulations for the growth of fastidious bacteria that require specific nutrients or additional growth factors. It provides a rich source of nitrogen, which is essential for microbial growth and protein synthesis. Casein hydrolysate can also be used as a substitute for complex peptones in certain applications, as it provides similar nutritional content.
Application
Casein hydrolysate has various applications in sterility testing media or biochemical characterization as it is utilized by a wide variety of microorganisms. It can also be used in the production of certain antibiotics, toxins, bacteriocins, and enzymes. It is especially suited for the large-scale cultivation of Diphteria bacteria, Tetanus bacilli and Streptococci. In industrial fermentations it favors a high yield of biomass, particularly for certain yeasts and fastidious bacilli.
Analysis Note
Appearance: Lightly yellow to lightly brownishyellow fine powder.
Identity (NIR): passes test
Total nitrogen (N) (Kjeldahl): 7.0 - 8.5 %
pH-value (5 %; water): 4.7 - 7.0
Amino nitrogen (as N): 5.0 - 6.5 %
Sulfated ash (800 °C): ≤ 58 %
Loss on Drying (105°C): ≤ 6.0 %
Suitability for microbiology: passes test
Identity (NIR): passes test
Total nitrogen (N) (Kjeldahl): 7.0 - 8.5 %
pH-value (5 %; water): 4.7 - 7.0
Amino nitrogen (as N): 5.0 - 6.5 %
Sulfated ash (800 °C): ≤ 58 %
Loss on Drying (105°C): ≤ 6.0 %
Suitability for microbiology: passes test
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.