Skip to Content
Merck
All Photos(1)

Key Documents

1.02003

Supelco

Cadmium acetate dihydrate

for analysis EMSURE®

Synonym(s):

Cadmium acetate dihydrate, Cadmium Acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Cd · 2H2O
CAS Number:
Molecular Weight:
266.53
MDL number:
UNSPSC Code:
12352103
EC Index Number:
208-853-2
NACRES:
NA.21
Assay:
≥99.0% ((CH3COO)2Cd.2H2O basis, complexometric)
form:
solid
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥99.0% ((CH3COO)2Cd.2H2O basis, complexometric)

form

solid

potency

333 mg/kg LD50, oral (Rat)

impurities

≤0.005% Insoluble matter
≤0.1% Substances not precipitated with H2S (as sulphates)

pH

7 (20 °C, 50 g/L in H2O)

mp

256 °C (anhydrous)

density

2.01 g/cm3 at 20 °C

bulk density

950 kg/m3

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
Na: ≤0.002%
Pb: ≤0.001%
Zn: ≤0.002%

storage temp.

2-30°C

SMILES string

[Cd](OC(=O)C)OC(=O)C.O.O

InChI

1S/2C2H4O2.Cd.2H2O/c2*1-2(3)4;;;/h2*1H3,(H,3,4);;2*1H2/q;;+2;;/p-2

InChI key

AUIZLSZEDUYGDE-UHFFFAOYSA-L

Related Categories

Analysis Note

Assay (komplexometric (CH₃COO)₂Cd·2H₂O): ≥ 99.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Sulfate (SO₄): ≤ 0.005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Na (Sodium): ≤ 0.002 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.002 %
Substances not precipitated with H₂S (as sulphates): ≤ 0.1 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - STOT RE 1 Oral

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service