Skip to Content

1.01883

Cellvento® Feed-200

Chemically defined cell culture feed for biopharmaceutical manufacturing in CHO Fed-batch, dry powder

Synonym(s):

CHO fed‑batch feed supplement, Cellvento Feed‑200, biopharmaceutical manufacturing feed, chemically defined CHO feed, dry powder CHO feed supplement, non-animal origin cell culture feed

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106213
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

non-animal source

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

technique(s)

cell culture | mammalian: suitable (suspension)

application(s)

pharma/biopharma processes

storage temp.

2-8°C

General description

Cellvento® Feed‑200 is a chemically defined, non‑animal origin cell culture feed designed for fed‑batch bioprocessing of Chinese Hamster Ovary (CHO) cells. It is supplied as a dry powder and, upon reconstitution, provides a concentrated blend of amino acids, vitamins, salts, and trace elements that replenish nutrients consumed during extended fed‑batch cultures. Used together with Cellvento® CHO‑200 production medium, Feed‑200 supports high‑density cell growth, extended culture duration, and improved recombinant protein / monoclonal antibody (mAb) titers in biotherapeutic development and manufacturing workflows.

Application

Cellvento® Feed‑200 is suitable for use in fed‑batch CHO cell culture processes to enhance cell growth, viability, and recombinant protein/mAb titers, including:

  • Companion feed for fed-batch CHO cell culture processes with Cellvento® CHO-200 medium.
  • Optimized for CHO-S cells (including GS expression systems); may also be suitable for other CHO cell lines.
  • Supports biotherapeutic manufacturing, including recombinant protein production, across research, process development, and manufacturing scales.
  • Enables feeding strategy optimization — including feed volume, frequency, and supplement levels — to enhance viable cell density, viability, and product titer throughout the fed-batch culture.

Features and Benefits

  • Chemically defined formulation – All components are of non-animal origin, eliminating undefined biological variability and supporting bioprocess development for biotherapeutic manufacturing.
  • Concentrated nutrient feed – Delivers amino acids, vitamins, salts, and trace elements to replenish depleted nutrients and sustain cell performance in extended fed-batch cultures.
  • Dry powder format: Easy to store and handle.
  • Optimized for CHO-S fed-batch processes – Designed to work with Cellvento® CHO-200 medium to support cell growth, viability, and recombinant protein productivity in biotherapeutic manufacturing.

Preparation Note

Formulated to contain 115.8 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.