Skip to Content
Merck

Skip To

1.01565

L-Asparagine monohydrate

EMPROVE® EXPERT, Ph. Eur., ChP

Synonym(s):

L-Asparagine monohydrate, Asparaginic acid semiamide, Asn

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
Molecular Weight:
150.14
UNSPSC Code:
12352209
EC Index Number:
200-735-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ChP, Ph. Eur.

description

EMPROVE® EXPERT Ph Eur

product line

EMPROVE® EXPERT

form

solid

pH

4.0-5.5 (20 °C, 20 g/L in H2O)

mp

215-217 °C (decomposes)

solubility

22 g/L

bulk density

530 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3][C@@H](CC(=O)N)C(=O)[O-].O

InChI

1S/C4H8N2O3.H2O/c5-2(4(8)9)1-3(6)7;/h2H,1,5H2,(H2,6,7)(H,8,9);1H2/t2-;/m0./s1

InChI key

RBMGJIZCEWRQES-DKWTVANSSA-N

General description

Our SAFC® portfolio of high-quality products for biopharmaceutical processing withstands strict quality control procedures and is produced according to MQ-500 requirements as defined by the M-Clarity program.

M-Clarity Program

As part of our EMPROVE® Program, our raw materials are offered with EMPROVE® Dossiers which provide comprehensive, up-to-date documentation to help you navigate regulatory challenges, manage risks, and improve your manufacturing processes.

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y00003051043513101566
description

EMPROVE® EXPERT, Ph. Eur., ChP

description

European Pharmacopoeia (EP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

for biochemistry

agency

ChP, Ph. Eur.

agency

-

agency

USP

agency

-

form

solid

form

-

form

-

form

solid

solubility

22 g/L

solubility

-

solubility

-

solubility

22 g/L

bulk density

530 kg/m3

bulk density

-

bulk density

-

bulk density

530 kg/m3

product line

EMPROVE® EXPERT

product line

-

product line

-

product line

-

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

-

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters