Skip to Content

1.01042

Aluminium potassium sulfate dodecahydrate

EMPROVE® ESSENTIAL, Ph. Eur., BP, USP

Synonym(s):

Aluminium potassium sulfate dodecahydrate, Potassium aluminium sulfate, Alum potassium, Potassium alum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
KAl(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
474.39
UNSPSC Code:
12352302
EC Index Number:
233-141-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, Ph. Eur., USP

product line

EMPROVE® ESSENTIAL

form

solid

pH

3.0-3.5 (20 °C, 100 g/L in H2O)

mp

92.5 °C

solubility

139 g/L

density

1.75 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-30°C

SMILES string

[K+].[Al]1(O[S](=O)(=O)O1)O[S](=O)(=O)[O-].O.O.O.O.O.O.O.O.O.O.O.O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Pharmaceutical grade aluminium potassium sulfate dodecahydrate is used in pharmaceutical industries as pH adjuster, buffer agents, in medicine mainly as an adstringent (or styptic) and antiseptic.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 1



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.