Skip to Content
Merck

1.01007

L-Alanine

for biochemistry

Synonym(s):

L-Alanine, 2-Aminopropionic acid, Ala

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-273-8
NACRES:
NA.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

pH

5.5-6.5 (20 °C, 100 g/L in H2O)

mp

295-297 °C (decomposition)

solubility

166.5 g/L

density

1.40 g/cm3 at 20 °C

bulk density

660 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]C(C)C(=O)[O-]

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)

InChI key

QNAYBMKLOCPYGJ-UHFFFAOYSA-N

General description

for biochemistry

Application

L-Alanine for biochemistry. CAS 56-41-7, pH 5.5 - 6.5 (100 g/l, H₂O, 20 °C).

Analysis Note

Assay (perchloric acid titration, calculated on dried substance)99.0 - 101.0 %Identity (IR-spectrum)passes testIdentity (specific rotation)passes testAppearancewhite to almost white crystalline powder or colorless crystalsAppearance of solution (25 g/l, water)clear and not more intense in color than reference solution BY₆Spec. rotation (α 20/D, 100 g/l, hydrochloric acid 250 g/l, calc. on dried substance)+13.5 to +15.5Chloride (Cl)≤ 200 ppmSulfate (SO₄)≤ 100 ppmAs (Arsenic)≤ 5 ppmCa (Calcium)≤ 50 ppmCo (Cobalt)≤ 5 ppmFe (Iron)≤ 5 ppmK (Potassium)≤ 5 ppmMg (Magnesium)≤ 5 ppmNa (Sodium)≤ 50 ppmPb (Lead)≤ 10 ppmZn (Zinc)≤ 5 ppmNinhydrin-positive substances (LC)(any ninhydrin-positive impurity)≤ 0.1 %Ninhydrin-positive substances (LC) (ammonium (570 nm))≤ 0.02 %Ninhydrin-positive substances (LC)(total impurities)≤ 0.5 %Loss on drying (105 °C; 3 h)
≤ 0.2 %≤ 0.2 %


Still not finding the right product?

Explore all of our products under L-Alanine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library