Skip to Content
Merck

Skip To

1.00366

(S)-Lactic acid

about 90%, EMPROVE® EXPERT, Ph. Eur., BP, E 270

Synonym(s):

L-(+)-Lactic acid, 2-Hydroxypropionic acid, Lactol, (S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
E Number:
E270
UNSPSC Code:
12352106
Beilstein/REAXYS Number:
1720251
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

BP, Ph. Eur.

vapor pressure

0.1 hPa ( 25 °C)

product line

EMPROVE® EXPERT

form

liquid

concentration

90%

technique(s)

API processing | cocrystal formation: suitable, API processing | salt formation: suitable

pH

2.8 (20 °C, 10 g/L in H2O)

bp

122 °C/20 hPa

mp

18 °C

density

1.21 g/cm3 at 20 °C

application(s)

liquid formulation
parenterals
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Lactic acid is used as an acidifying agent in a oral, topical and parenteral formulations.
Due to its low microbial and endotoxin limits, (S)-Lactic acid about 90% Emprove® Expert is especially suitable for high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1356734PHR3879L1750
concentration

90%

concentration

-

concentration

-

concentration

-

application(s)

liquid formulation
parenterals
pharmaceutical
semi-solid formulation

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

-

agency

BP, Ph. Eur.

agency

-

agency

traceable to USP 1356734

agency

-

technique(s)

API processing | cocrystal formation: suitable, API processing | salt formation: suitable

technique(s)

HPLC: suitable

technique(s)

-

technique(s)

-

form

liquid

form

-

form

-

form

-

density

1.21 g/cm3 at 20 °C

density

-

density

-

density

-


Still not finding the right product?

Explore all of our products under (S)-Lactic acid


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters