Skip to Content
Merck

Skip To

1.00201

Phenol

EMPROVE® ESSENTIAL, Ph. Eur., ChP, JP, USP

Synonym(s):

Hydroxybenzene, Carbolic acid, Hydroxybenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
EC Index Number:
203-632-7
UNSPSC Code:
12352104
Beilstein/REAXYS Number:
969616
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

ChP, JP, Ph. Eur., USP

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C), 0.36 mmHg ( 20 °C)

product line

EMPROVE® ESSENTIAL

assay

99.0-100.5% (bromatometric)

form

solid

autoignition temp.

1319 °F

expl. lim.

8.6 %

technique(s)

API processing | cocrystal formation: suitable

pH

5 (20 °C, 50 g/L in H2O)

bp

182 °C (lit.)

mp

40-42 °C (lit.)

transition temp

flash point 81 °C

density

1.071 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Phenol is active against a wide range of microorganisms and is therefore used as a preservative in pharmaceutical formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA′s Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada’s Pest Management Regulatory Agency, Turkey’s Biocidal Products Regulation, Korea’s Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1047152480635952
description

EMPROVE® ESSENTIAL, Ph. Eur., ChP, JP, USP

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

United States Pharmacopeia (USP) Reference Standard

description

PESTANAL®, analytical standard

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

agriculture
cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

technique(s)

API processing | cocrystal formation: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

agency

ChP, Ph. Eur., JP, USP

agency

traceable to USP 1524806

agency

USP

agency

-

form

solid

form

-

form

-

form

-

assay

99.0-100.5% (bromatometric)

assay

-

assay

-

assay

-

product line

EMPROVE® ESSENTIAL

product line

-

product line

-

product line

PESTANAL®


Still not finding the right product?

Explore all of our products under Phenol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Nervous system,Kidney,Liver,Skin

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

177.8 °F - closed cup

flash_point_c

81 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difficulty Finding Your Product Or Lot/Batch Number?

Product numbers are combined with Pack Sizes/Quantity when displayed on the website (example: T1503-25G). Please make sure you enter ONLY the product number in the Product Number field (example: T1503).

Example:

T1503
Product Number
-
25G
Pack Size/Quantity

Additional examples:

705578-5MG-PW

PL860-CGA/SHF-1EA

MMYOMAG-74K-13

1000309185

enter as 1.000309185)

Having trouble? Feel free to contact Technical Service for assistance.



Questions

Reviews

No rating value

Active Filters