Skip to Content
Merck

Skip To

1.00181

Bactident Oxidase

Cytochrome Oxidase Test Strips, 50 strips/pack

Synonym(s):

Cytochrome Oxidase Test Strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.84
UNSPSC Code:
41116130
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

manufacturer/tradename

Bactident

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

storage temp.

2-8°C

suitability

selective and differential for

General description

Bactident Oxidase test strips are designed to detect cytochrome oxidase positive microorganisms in a test sample or grown culture.
Cytochrome oxidase is an enzyme found in specific microbes which will catalyze the substrate cytochrome c indicated by a color change of blue to blue violet. This reaction can be used to classify and identify the bacteria.
In the presence of molecular oxygen, the cytochrome oxidase/cytochrome c system can reduce all the available organic substances in the environment e.g. NaDi (1-naphthol dimethylparaphenylene diamine) reagent with the formation of condensation molecule indophenol blue.

Application

Bactident Oxidase test strips are used to detect cultured microorganisms which can catalyze cytochrome c substrate through a color change reaction.

Legal Information

BACTIDENT is a trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
405601.133011.13303
technique(s)

microbiological culture: suitable

technique(s)

microbe id | specific enzyme detection: suitable

technique(s)

microbe id | specific enzyme detection: suitable

technique(s)

microbe id | specific enzyme detection: suitable

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

application(s)

food and beverages
pharmaceutical

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical, microbiology

application(s)

detection, environmental
food and beverages

application(s)

detection, environmental
food and beverages

form

solid

form

-

form

-

form

-

suitability

selective and differential for

suitability

Plesiomonas spp., Aeromonas spp., Neisseria spp., Pseudomonas spp.

suitability

Aeromonas spp., Bacillus spp., Salmonella spp.

suitability

Citrobacter spp., Escherichia coli, Proteus spp.

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters