1.00136
Benzoic acid
for analysis EMSURE® ACS, Reag. Ph Eur
Synonym(s):
Benzoic acid, Phenylformic acid, Benzene carboxylic acid
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. Ph. Eur.
vapor pressure
0.001 hPa ( 20 °C)
product line
EMSURE®
Assay
≥99.9% (acidimetric)
form
solid
autoignition temp.
570 °C
potency
1700 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)
impurities
≤0.002% Sulfur compounds (as S)
≤0.005% In methanol insoluble matter
≤0.5% Water
sulfated ash
≤0.005%
pH
2.5-3.5 (20 °C in H2O, saturated solution)
bp
249 °C/1013 hPa
mp
121-123 °C
transition temp
flash point 121 °C
solubility
2.9 g/L
density
1.321 g/cm3 at 20 °C
bulk density
500 kg/m3
anion traces
other halogens (as Cl-): ≤0.005%
sulfate (SO42-): ≤0.002%
cation traces
Fe: ≤0.005%
heavy metals (as Pb): ≤0.0005%
storage temp.
2-30°C
SMILES string
[O-]C(=O)c1ccccc1.[H+]
InChI
1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)
InChI key
WPYMKLBDIGXBTP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Phytochemical Composition and Antimicrobial Efficacy of Salvadora persica Root Extracts Against Carbapenem-Resistant Acinetobacter baumannii.: The study examines the antimicrobial efficacy of Salvadora persica root extracts, highlighting the role of benzoic acid in the phytochemical composition. (Wan Alias et al., 2024).
- Promoting Water Oxidation by Proton Acceptable Groups Surrounding Catalyst on Electrode Surface.: This research explores the enhancement of water oxidation processes using benzoic acid derivatives as proton acceptor groups on electrode surfaces. (Li et al., 2024).
Analysis Note
Identity: passes test
Appearance of solution (5 %; ethanol 96 %): passes test
Appearance of solution (color): passes test
Appearance of solution (clearness): passes test
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.0005 %
Fe (Iron): ≤ 0.005 %
halogenated compounds (as Cl): ≤ 0.005 %
Sulfur compounds (as S): ≤ 0.002 %
In methanol insoluble matter: ≤ 0.005 %
Oxidizable matter: passes test
Substances reducing permanganate: passes test
Readily carbonisable substance: passes test
Sulfated ash: ≤ 0.005 %
Water: ≤ 0.5 %
Corresponds to Reag. Ph Eur
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Summary application report for analysis of moisture in 2-Methylpiperidine
Accurately measure the moisture content in 2-Propylamine ((CH₃)₂CHNH₂) through Karl Fischer titration, using both volumetric and coulometric methods.
Accurately measure the moisture content in 2-Propylamine ((CH₃)₂CHNH₂) through Karl Fischer titration, using both volumetric and coulometric methods.
Accurately measure the moisture content in 2-Propylamine ((CH₃)₂CHNH₂) through Karl Fischer titration, using both volumetric and coulometric methods.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service