Skip to Content
Merck
All Photos(3)

Key Documents

05-515

Sigma-Aldrich

Anti-acetyl-Lysine Antibody, clone 4G12

clone 4G12, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
4G12, monoclonal
application:
IP
WB
species reactivity:
human, mouse, rat, vertebrates
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
68

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G12, monoclonal

species reactivity

human, mouse, rat, vertebrates

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

General description

In the nucleus, DNA is tightly packed into nucleosomes generating an environment which is highly repressive towards DNA processes such as transcription. Acetylation of lysine residues within proteins has emerged as an important mechanism used by cells to overcome this repression. The acetylation of non-histone proteins such as transcription factors, as well as histones appears to be involved in this process. Acetylation may result in structural transitions as well as specific signaling within discrete chromatin domains. The role of acetylation in intracellular signaling has been inferred from the binding of acetylated peptides by the conserved bromodomain. Furthermore, recent findings suggest that bromodomain/acetylated-lysine recognition can serve as a regulatory mechanism in protein-protein interactions in numerous cellular processes such as chromatin remodeling and transcriptional activation.

Specificity

Acetylated lysine-containing proteins including histones. Of all the possible H3 and H4 acetylation sites, this antibody exhibits highest affinity for histone H4 acetylated on lysine 8 and histone H3 acetylated on lysine 14.
Expected to cross-react with rat and mouse.

Immunogen

Mixture of chemically acetylated antigens; recognizes numerous proteins acetylated on a lysine, most prominently on histones.

Application

Detect acetyl-Lysine with Anti-acetyl-Lysine Antibody, clone 4G12 (Mouse Monoclonal Antibody), that has been shown to work in IP & WB.
Immunoprecipitation:
5 μg of a previous lot immunoprecipitated in vitro acetylated PCAF added to a 3T3 RIPA cell lysate. The immunoprecipitated PCAF was detected by subsequent western blot analysis using 1 μg/mL monoclonal anti-GST (Catalog # 05-311).

Quality

Routinely evaluated by Western Blot on untreated and sodium butyrated treated HeLa lysates.

Western Blot Analysis:
1:500 dilution of this lot detected ACETYL-LYSINE on 10 μg of sodium butyrated treated HeLa lysates.

Target description

Varies depending upon the protein being detected.

Physical form

Format: Purified
Purified mouse monoclonal IgG in buffer containing 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide, before the addition of glycerol to 30%.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contrasting proteome biology and functional heterogeneity of the 20 S proteasome complexes in mammalian tissues.
Gomes, AV; Young, GW; Wang, Y; Zong, C; Eghbali, M; Drews, O; Lu, H; Stefani, E; Ping, P
Molecular and Cellular Proteomics null
Jasmyne Carnevale et al.
Molecular and cellular biology, 32(5), 900-912 (2011-12-21)
E2F transcription can lead to cell proliferation or apoptosis, indicating that E2Fs control opposing functions. In a similar manner, DNA double-strand breaks can signal to induce cell cycle arrest or apoptosis. Specifically, pRB is activated following DNA damage, allowing it
Wai Chi Ho et al.
Cancer research, 65(10), 4273-4281 (2005-05-19)
The primary goal of chemotherapy is to cause cancer cell death. However, a side effect of many commonly used chemotherapeutic drugs is the activation of nuclear factor-kappaB (NF-kappaB), a potent inducer of antiapoptotic genes, which may blunt the therapeutic efficacy
Natsuko Izumi et al.
Science signaling, 3(116), ra27-ra27 (2010-04-08)
Phosphatidylinositol 3-kinase-related protein kinase (PIKK) family proteins play essential roles in DNA-based and RNA-based processes, such as the response to DNA damage, messenger RNA (mRNA) quality control, transcription, and translation, where they contribute to the maintenance of genome integrity and
Acetylation of general transcription factors by histone acetyltransferases.
Imhof, A, et al.
Current Biology, 7, 689-692 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service