Skip to Content
Merck
All Photos(1)

Key Documents

05-457

Sigma-Aldrich

Anti-Histone H1 Antibody, clone AE-4

clone AE-4, Upstate®, from mouse

Synonym(s):

H1 histone family, member 4, Histone H1b, histone 1, H1e, histone cluster 1, H1e

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AE-4, monoclonal

species reactivity

bovine, human, plant

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... H1-4(617854)
human ... H1-4(3008)

General description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures.

Specificity

Predicted to stain nuclei from other animal and plant species.
This antibody recognizes histone H1, Mr 32-33 kDa. Doublet detected due to hyperphosphorylation of histone H1 subpopulation. Cross-reacts with other histone isoforms in the following order of signal strength: H3, H2B > H4 > H2A, as determined with recombinant protein.

Immunogen

Nuclei from myeloid leukemia biopsy cells. Clone AE-4.

Application

Immunocytochemistry:
5-20 µg/mL of a previous lot showed positive nuclear immunostaining in A431 cells fixed with 4% paraformaldehyde and methanol.

ELISA:
Recommended.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-Histone H1 Antibody, clone AE-4 (mouse monoclonal antibody) validated in ELISA, ICC, WB to detect Histone H1 also known as H1 histone family member 4, Histone H1b, histone 1 H1e, histone cluster 1 H1e.

Quality

Routinely evaluated by immunoblot on lysates from an acid extract of HeLa cells.

Western Blot:
1-4 µg/mL of this lot detected histone H1 in lysates from an acid extract of HeLa cells.

Target description

32-33 kDa (Histone H1); Doublet (hyperphosphorylated Histone H1); 17 kDa (other histone isoforms).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2a in buffer containing PBS, pH 7.2, with 0.1% sodium azide. Frozen solution.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Nuclear extracts from NIH3T3 or HeLa cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicotine induces chromatin remodelling through decreases in the methyltransferases GLP, G9a, Setdb1 and levels of H3K9me2.
Chase, KA; Sharma, RP
The International Journal of Neuropsychopharmacology null
Super-resolution microscopy reveals decondensed chromatin structure at transcription sites.
Wang, Y; Maharana, S; Wang, MD; Shivashankar, GV
Scientific Reports null
Kwang-Hee Lee et al.
The Plant cell, 23(12), 4298-4317 (2011-12-14)
The regulatory particle (RP) of the 26S proteasome contains a heterohexameric ring of AAA-ATPases (RPT1-6) that unfolds and inserts substrates into the core protease (CP) for degradation. Through genetic analysis of the Arabidopsis thaliana gene pair encoding RPT2, we show
Jung Ha Choo et al.
BMC genomics, 8, 479-479 (2008-01-02)
MacroH2A1 is a histone variant that is closely associated with the repressed regions of chromosomes. A recent study revealed that this histone variant is highly enriched in the inactive alleles of Imprinting Control Regions (ICRs). The current study investigates the
Jae Hyun Lee et al.
Human molecular genetics, 18(14), 2567-2574 (2009-04-22)
We recently described two opposing states of transcriptional competency. One is termed 'competent' whereby a gene is capable of responding to trans-acting transcription factors of the cell, such that it is active if appropriate transcriptional activators are present, though it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service