Skip to Content
Merck
All Photos(1)

Key Documents

05-368MG

Sigma-Aldrich

Anti-phospho-Ser/Thr-Pro MPM-2 Antibody

clone MPM-2, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MPM-2, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

Gene Information

human ... FOXM1(2305)

General description

This antibody recognizes a variety of proteins that are phosphorylated during mitosis.
Upon entry into M (Mitosis) phase, many proteins are phosphorylated either directly or indirectly by M-phase-promoting factor (MPF). The MPM2 monoclonal antibody binds to a phospho amino acid-containing epitope (peptides containing LTPLK and FTPLQ domains) present on more than 50 proteins of M-phase eukaryotic cells.

Specificity

Recognizes phosphorylated serines or threonines when followed by a proline.

Immunogen

Mitotic human HeLa cell cytosolic lysate

Application

Detect phospho-Ser/Thr-Pro MPM-2 also known as Mitotic protein #2 with Anti-phospho-Ser/Thr-Pro MPM-2 Antibody (Mouse Monoclonal Antibody), that has been demonstrated to work in ELISA, IHC, IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

routinely evaluated in immunoblot on colcemid treated human HeLa carcinoma cells

Target description

varies depending on the protein detected

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in mouse IgG in 1.0 mL PBS containing no preservatives.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Colcemid-treated HeLa (lane 2) cell lysates, A431 mitotic cells fixed with 3.7% paraformaldehyde

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rong Sang et al.
Cell & bioscience, 12(1), 78-78 (2022-06-02)
Mental retardation is a complex neurodevelopmental disorder. NPAT, a component of the histone locus body (HLB), has been implicated as a candidate gene for mental retardation, with a mechanism yet to be elucidated. We identified that mxc, the Drosophila ortholog
Phosphoproteins are components of mitotic microtubule organizing centers.
Vandre, D D, et al.
Proceedings of the National Academy of Sciences of the USA, 81, 4439-4443 (1984)
Sequence-specific and phosphorylation-dependent proline isomerization: a potential mitotic regulatory mechanism.
Yaffe, M B, et al.
Science (New York, N.Y.), 278, 1957-1960 (1997)
Mitosis-specific monoclonal antibody MPM-2 inhibits Xenopus oocyte maturation and depletes maturation-promoting activity.
Kuang, J, et al.
Proceedings of the National Academy of Sciences of the USA, 86, 4982-4986 (1989)
Makoto T Hayashi et al.
Nature, 522(7557), 492-496 (2015-06-26)
Tumour formation is blocked by two barriers: replicative senescence and crisis. Senescence is triggered by short telomeres and is bypassed by disruption of tumour-suppressive pathways. After senescence bypass, cells undergo crisis, during which almost all of the cells in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service