Skip to Content

05-166

Anti-IGF-II Antibody, clone S1F2

clone S1F2, Upstate®, from mouse

Synonym(s):

Anti-C11orf43, Anti-GRDF, Anti-IGF-II, Anti-PP9974, Anti-SRS3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
S1F2, monoclonal
Application:
ELISA, ICC, IP, Neutral, WB
Citations:
44
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

S1F2, monoclonal

species reactivity

human, rat

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, neutralization: suitable, western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IGF2(3481)

General description

Insulin-like growth factor 2 (UniProt: P01346; also known Insulin-like growth factor II, IGF-II, Multiplication-stimulating activity, MSA, Multiplication-stimulating polypeptide) is encoded by the Igf2 (also known Igf-2) gene (Gene ID: 24483) in rat. This gene encodes a member of the insulin family of polypeptide growth factors that is primarily involved in fetal development, influencing cellular proliferation and differentiation. It is an imprinted gene, with expression predominantly from the paternally inherited allele. It is regulated by multiple epigenetic mechanisms, including differential methylation and transcription factor binding at the IGF2-H19 locus on chromosome 11p15. It is a candidate gene for eating disorders. IGF-II is a widely expressed, secreted protein that is synthesized with a signal peptide (aa 1-24), propeptide (aa 92-180) which is subsequently cleaved off to produce the mature protein (aa 25-91). Human IGF-II shares similar structural features with IGFI, including a 62% sequence homology. Alternatively spliced transcript variants, encoding either the same or different isoform, have been found for this gene. In human plasma, IGFI and IGFII are associated with IGF-binding proteins that transport the polypeptides and partially regulate their actions in vivo. In addition to the insulin receptor, IGFII binds to two forms of IGF receptors, both of which are widely distributed in different tissues and cultured cells. IGFII is mitogenic for a variety of cultured cells, including mouse 3T3 cells,12 normal rat kidney cells, human or chicken fibroblasts and MCF-7 human breast carcinoma cells. Dysregulation of IGF-II expression has been implicated in various diseases, including overgrowth syndromes such as Beckwith-Wiedemann syndrome and Silver-Russell syndrome, as well as in several cancers, where IGF-II promotes tumor growth and survival. (Ref.: Wilkin, F., et al. (2000). J Clin Endocrinol Metab. 85(5):2048-56; Boo, H-J., et al. (2016). Nat Commun. 7:12961; Mellott, T.J., et al. (2017). PLoS One. 12(1):e0170450; Zolov, S.N., et al. (2021). J Biol Chem. 296:100712; Birzniece, V., et al. (2022). Endocr Connect. 11(4):e210375).
~7 kDa

Immunogen

Partially purified rat IGF-II.

Application

Anti-IGF-II, clone S1F2, Cat. No. 05-166, is a mouse monoclonal antibody that detects IGF-II and is tested for use in Western Blotting and Immunohistochemistry.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling
Tested Applications

Western Blotting Analysis: A representative lot detected IGF-II in Western Blotting applica tion (Mellott, T.J., et al. (2017). PLoS One.12(1):e0170450; Zolov, S.N., et al. (2021). J Biol Chem.296:100712).

Immunohistochemistry Analysis: A representative lot detected IGF-II in Immunohistochemistry application (Wilkin, F., et al. (2000). J Clin Endocrinol Metab. 85(5):2048-56; Boo, H-J., et al. (2016). Nat Commun. 7:12961).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone S1F2 is a mouse monoclonal antibody that specifically detects IGF-II. This antibody has less than 10% cross-reactivity to human IGF-I.

Physical form

Format: Purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Recommended storage: +2°C to +8°C.

Analysis Note

Evaluated by Western Blotting with recombinant human IGF-II.

Western Blotting Analysis (WB): A 1:1000 dilution of this antibody detected recombinant human IGF-II.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library