Skip to Content
Merck
All Photos(4)

Key Documents

05-1312-I

Sigma-Aldrich

Anti-Ubiquityl-Histone H2B Antibody, clone 56

clone 56, from mouse

Synonym(s):

Histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, H2B/al, Histone H2B.g, H2B/g, Histone H2B.h, H2B/h, Histone H2B.k, H2B/k, Histone H2B.l, H2B/l)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

56, monoclonal

species reactivity

mouse, human, rat

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

General description

Histone H2B type 1-C/E/F/G/I (UniProt: P62807; also known as Histone H2B.1 A, Histone H2B.a, H2B/a, Histone H2B.g, H2B/g, Histone H2B.h, H2B/h, Histone H2B.k, H2B/k, Histone H2B.l, H2B/l) is encoded by the HIST1H2BC (also known as H2BFL, HIST1H2BE H2BFH, HIST1H2BF, H2BFG, HIST1H2BG, H2BFA, HIST1H2BI, H2BFK) gene (Gene ID: 3017, 8339, 8343, 8344, 8346, 8347) in human. Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. Histone modifications regulate DNA transcription, repair, recombination, and replication. Histone H2B is one of the core components of the nucleosome, the smallest subunit of chromatin. DNA base pairs are wrapped around an octamer of core histone proteins consisting of two each of Histone H2A, H2B, H3 and H34. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. A number of studies have implicated ubiquitinated H2B in heterochromatic gene silencing and in the control of gene expression. Histone H2B is ubiquitylated at Lys120 by RAD6 E2 protein in conjunction with RNF20/RNF40 and this ubiquitinylated form is found preferentially associated with the transcribed regions of highly expressed genes.

Specificity

Clone 56 recognizes histone H2B when ubiquitinated on Lys120. It does not recognize H2B in its unmodified state or ubiquitin unconjugated to H2B.

Immunogen

A branched peptide corresponding to the ubiquitination site on human H2B.

Application

Anti-Ubiquityl-Histone H2B, clone 56 Antibody, Cat. No. 05-1312-I, is a highly specific mouse monoclonal antibody that targets Ubiquityl-Histone H2B and has been tested in Chromatin Immunoprecipitation (ChIP) and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected Ubiquityl-Histone H2B in HeLa, NIH3T3, and PC-12 cell lysate.

Western Blotting Analysis: A representative lot detected Ubiquityl-Histone H2B in U2OS whole cell extract, purified histone H2A, purified H2B, and HEK293 acid extracted proteins (Courtesy of Dr. Moshe Oren, Weizmann Institute of Science).

Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected Ubiquityl-Histone H2B in uH2B and H3K4me2 (Courtesy of Dr. Moshe Oren, Weizmann Institute of Science).

Quality

Evaluated by Western Blotting in PC-12 cell lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected Ubiquityl-Histone H2B in 10 µg of PC-12 cell lysate.

Target description

~25 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Smaragda Fanourgakis et al.
Computational and structural biotechnology journal, 21, 614-629 (2023-01-21)
The timing and location of writing and erasing of histone modifications determine gene expression programs and are tightly controlled processes. One such modification is the monoubiquitination of histone H2B (H2Bub), whose precise level during transcription elongation is dynamically regulated by
Jennifer J Chen et al.
Molecular and cellular biology, 40(15) (2020-05-06)
Rtf1 is a conserved RNA polymerase II (RNAPII) elongation factor that promotes cotranscriptional histone modification, RNAPII transcript elongation, and mRNA processing. Rtf1 function requires the phosphorylation of Spt5, an essential RNAPII processivity factor. Spt5 is phosphorylated within its C-terminal domain
Zuntao Xu et al.
The Plant journal : for cell and molecular biology, 110(6), 1670-1680 (2022-04-09)
Heading date is one of the most pivotal agronomic traits for rice (Oryza sativa) yield and adaptation. Little is known about the crosstalk between histone ubiquitination and histone methylation in rice heading date regulation. Here, we reported HISTONE MONOUBIQUITINATION 1
Bei Bu et al.
The EMBO journal, 39(1), e101259-e101259 (2019-09-21)
Psychiatric diseases are often accompanied by circadian disruptions, but the molecular underpinnings remain largely unclear. To address this, we screened genes that have been previously reported to be associated with psychiatric diseases and found that TRRAP, a gene associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service