Skip to Content
Merck
All Photos(1)

Key Documents

N-912

Supelco

Norbuprenorphine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H35NO4
CAS Number:
Molecular Weight:
413.55
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4)c56

InChI

1S/C25H35NO4/c1-21(2,3)22(4,28)16-13-23-8-9-25(16,29-5)20-24(23)10-11-26-17(23)12-14-6-7-15(27)19(30-20)18(14)24/h6-7,16-17,20,26-28H,8-13H2,1-5H3/t16-,17-,20-,22+,23-,24+,25-/m1/s1

InChI key

YOYLLRBMGQRFTN-IOMBULRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Norbuprenorphine is the primary active metabolite of the opiate buprenorphine in urine. This Certified Spiking Solution® is suitable as a starting material for use as calibrators and controls in GC/MS and LC/MS methods in forensic analysis, clinical toxicology, urine drug testing, or pain prescription monitoring. Buprenorphine, sold under trade names such as Buprenex®, Subutex® and Suboxone®, is an opiate drug used to treat opioid addiction and to moderate acute pain.

Legal Information

Buprenex is a registered trademark of Reckitt & Colman (Overseas) Ltd
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Suboxone is a registered trademark of RB Pharmaceuticals Limited
Subutex is a registered trademark of RB Pharmaceuticals Limited

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jinda Fan et al.
Bioconjugate chemistry, 22(4), 752-758 (2011-03-26)
Norbuprenorphine-3-β-d-glucuronide (nBPN-3-β-d-G, 1) is a major phase II metabolite of buprenorphine, a pharmaceutical used for the treatment of opioid addiction. The pharmacological activity of compound 1 is not clear because investigations have been limited by the lack of chemically pure
Panagiota Nikolaou et al.
Biomedical chromatography : BMC, 26(3), 358-362 (2011-07-02)
Buprenorphine (BUP) is used for the maintenance of opioid-addicted pregnant women. Because BUP and its main metabolite nor-BUP are excreted into breast milk, a sensitive and specific GC/MS method has been developed, optimized and validated for their determination in breast
Xia Luo et al.
Antimicrobial agents and chemotherapy, 56(7), 3641-3647 (2012-05-09)
This was an open-label, single-sequence trial in hepatitis C virus-negative volunteers on stable, individualized, buprenorphine maintenance therapy. Telaprevir at 750 mg every 8 h was coadministered with buprenorphine/naloxone (4:1 ratio as sublingual tablets) for 7 days with food. Pharmacokinetic profiles
Ioannis I Papoutsis et al.
Journal of pharmaceutical and biomedical analysis, 54(3), 588-591 (2010-10-14)
A sensitive and specific GC/MS method for the determination of buprenorphine (BPN) and its main metabolite nor-buprenorphine (nor-BPN) in blood has been developed, optimized and validated. Sample preparation includes solid-phase extraction of both analytes and their derivatization with acetic anhydride
Sarah M Brown et al.
Anesthesiology, 115(6), 1251-1260 (2011-11-01)
The long-lasting high-affinity opioid buprenorphine has complex pharmacology, including ceiling effects with respect to analgesia and respiratory depression. Plasma concentrations of the major buprenorphine metabolites norbuprenorphine, buprenorphine-3-glucuronide, and norbuprenorphine-3-glucuronide approximate or exceed those of the parent drug. Buprenorphine glucuronide metabolites

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service