Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)
concentration
1.0 mg/mL in methanol
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
2-8°C
SMILES string
[2H]C([2H])([2H])NC(C([2H])([2H])[2H])([2H])C([H])([2H])C1=C([H])C([H])=C([H])C([H])=C1[H]
InChI
1S/C10H15N/c1-9(11-2)8-10-6-4-3-5-7-10/h3-7,9,11H,8H2,1-2H3/i1D3,2D3,8D,9D
InChI key
MYWUZJCMWCOHBA-URFKORASSA-N
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) forensics and toxicology | application(s) forensics and toxicology | application(s) forensics and toxicology | application(s) forensics and toxicology |
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| form liquid | form liquid | form liquid | form liquid |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| packaging ampule of 1 mL | packaging ampule of 1 mL | packaging ampule of 1 mL | packaging ampule of 1 mL |
Still not finding the right product?
Explore all of our products under (±)-Methamphetamine-D8 solution
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




