Skip to Content
Merck
All Photos(1)

Key Documents

L-001

Supelco

LSD solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Lysergic acid diethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H25N3O
CAS Number:
Molecular Weight:
323.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

O=C(N(CC)CC)[C@@]1([H])CN(C)[C@]2([H])CC3=CNC4=CC=CC(C2=C1)=C43

InChI

1S/C20H25N3O/c1-4-23(5-2)20(24)14-9-16-15-7-6-8-17-19(15)13(11-21-17)10-18(16)22(3)12-14/h6-9,11,14,18,21H,4-5,10,12H2,1-3H3/t14-,18-/m1/s1

InChI key

VAYOSLLFUXYJDT-RDTXWAMCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LSD, or lysergic acid diethylamide, is an illicit psychedelic drug with potent hallucinogenic effects such as altered thinking processes, closed- and open-eye visuals, synesthesia and an altered sense of time. This Snap-N-Spike® reference solution is suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

George Harrison's inner light.
Guilherme Nogueira M de Oliveira et al.
Revista brasileira de psiquiatria (Sao Paulo, Brazil : 1999), 34(4), 517-518 (2013-02-23)
Viola Fahmy et al.
Age and ageing, 41(4), 553-556 (2012-04-12)
little is known about illicit drug use in older people. Prevalences may rise as populations for whom illicit drug use has been more common and acceptable become older. to quantify illicit drug use in people aged 50 years and over
Alexander Paulke et al.
Analytical and bioanalytical chemistry, 404(2), 531-538 (2012-06-15)
The ergot alkaloid lysergic acid amide (LSA) is a secondary plant constituent in a number of plants, but it is mainly present in considerable amounts in Convolvulaceae, like Argyreia nervosa. Due to its close structural similarity to lysergic acid diethylamide
Theresa M Carbonaro et al.
Psychopharmacology, 226(2), 241-246 (2012-10-17)
Serotonergic hallucinogens such as (+)-lysergic acid diethylamide (LSD) and dimethyltryptamine (DMT) produce distinctive visual effects, whereas the synthetic hallucinogen N,N-diisopropyltryptamine (DiPT) is known for its production of auditory distortions. This study compares the discriminative stimulus effects of DiPT to those
Jufang Shan et al.
PLoS computational biology, 8(4), e1002473-e1002473 (2012-04-26)
From computational simulations of a serotonin 2A receptor (5-HT(2A)R) model complexed with pharmacologically and structurally diverse ligands we identify different conformational states and dynamics adopted by the receptor bound to the full agonist 5-HT, the partial agonist LSD, and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service