Skip to Content
Merck
All Photos(1)

Key Documents

H-076

Supelco

9-Hydroxyrisperidone solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27FN4O3
CAS Number:
Molecular Weight:
426.48
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC1=C(CCN2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5CCCC(O)C5=N1

InChI

1S/C23H27FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h4-5,13,15,19,29H,2-3,6-12H2,1H3

InChI key

PMXMIIMHBWHSKN-UHFFFAOYSA-N

Gene Information

General description

A Snap-N-Spike® Reference Solution suitable to use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, and forensic analysis. Also known as paliperidone, 9-Hydroxyrisperidone is sold under the brand name Invega, an atypical antipsychotic used for acute and maintenance treatment of schizophrenia. The drug is also the primary metabolite of risperidone, marketed as the atypical antipsychotic Risperdal®.

Application


  • 9-HydroxyrisperiDonesolution for in vitro research: Employed in pharmacological studies to investigate dual antipsychotic therapies in treatment-resistant conditions, enhancing the understanding of drug interactions and efficacy (Warnick et al., 2024).

  • High-purity 9-HydroxyrisperiDoneliquid for pharmacological studies: Utilized in crystallography research to assess the impact of alkyl chain length on the crystallization behavior of related compounds, providing insights into drug formulation strategies (Chen et al., 2024).

  • Buy 9-HydroxyrisperiDonesolution for receptor binding assays: Applied in the development of novel drug delivery systems such as dissolving microneedles, enhancing the administration of long-acting antipsychotic treatments (Londhe and Bhadale, 2023).

  • 9-HydroxyrisperiDoneactive metabolite for antipsychotic research: Analyzed in postmortem toxicological assessments to identify metabolites, improving forensic analysis techniques and therapeutic drug monitoring (Yamagishi et al., 2023).

  • Analytical standard 9-HydroxyrisperiDonefor biochemistry labs: Investigated in the synthesis of prodrug microcrystals for extended-release formulations, aiding in the design of more effective and patient-friendly antipsychotic medications (Xu et al., 2022).




Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Risperdal is a registered trademark of Johnson & Johnson
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Yvette Roke et al.
Journal of child and adolescent psychopharmacology, 22(6), 432-439 (2012-12-14)
The aim of this study was to investigate the long-term treatment effects of risperidone on prolactin levels and prolactin-related side effects in pubertal boys with autism spectrum disorders (ASD) and disruptive behavior disorders (DBD). Physical healthy 10-20-year-old males with ASD
Pierre S Chue et al.
Expert review of neurotherapeutics, 12(12), 1399-1410 (2012-12-15)
Paliperidone, or 9-hydroxyrisperidone (Invega(®), Janssen, Antwerp, Belgium) is the major active metabolite of the atypical antipsychotic risperidone (Risperdal(®), Janssen). It possesses a similar, though not identical, receptor pharmacology to the parent molecule. There are additional differences in terms of its
Danielle M Butzbach et al.
Journal of forensic sciences, 58(1), 90-100 (2012-09-22)
The stability of two benzisoxazole antipsychotics was determined in vitro in decomposing porcine blood inoculated with bacteria, utilizing a high-performance liquid chromatography with ultraviolet and fluorescence detection method for drug quantitation. Stability experiments for risperidone and paliperidone were conducted at
Drug and therapeutics bulletin, 50(9), 102-105 (2012-09-12)
For over 40 years, antipsychotic drugs have been used as long-term maintenance treatment to control symptoms and reduce relapse rates in patients with schizophrenia. 'First-generation' oral agents such as haloperidol and chlorpromazine are associated with high levels of unwanted neurological
Paliperidone as an alternative for risperidone in a case of schizophrenia with retrograde ejaculation.
Arun Kandasamy
The Journal of neuropsychiatry and clinical neurosciences, 24(3), E17-E18 (2012-10-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service