Skip to Content
Merck
All Photos(1)

Documents

E-043

Supelco

Ethanol-20 (5 ampules/kit)

20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®

Synonym(s):

Ethanol solution, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 5 × 5 mL

manufacturer/tradename

Cerilliant®

concentration

20 mg/dL in H2O

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A kit of NIST traceable ethanol standards suitable for calibration of ethanol testing methods. These Certified Reference Materials are widely used in forensic analysis and clinical toxicology applications for determination of blood alcohol content.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Enocksson et al.
The Journal of clinical investigation, 95(5), 2239-2245 (1995-05-01)
Although it is well established in several mammalian species that beta 3-adrenoceptors play a major role in regulating lipolysis and thermogenesis in adipose tissue, the functional existence and role of this receptor subtype in man has been controversial. We investigated
Thiago M Pais et al.
PLoS genetics, 9(6), e1003548-e1003548 (2013-06-12)
The yeast Saccharomyces cerevisiae is able to accumulate ≥17% ethanol (v/v) by fermentation in the absence of cell proliferation. The genetic basis of this unique capacity is unknown. Up to now, all research has focused on tolerance of yeast cell
J Piskur et al.
FEBS letters, 375(3), 174-178 (1995-11-20)
A recently developed mechanochemical method has provided a new, efficient tool for studies on the thermal stability and structure of aggregated DNA in ethanol-water solutions. At low ethanol concentrations DNA is fully soluble and is in the B form. However
A Nyéki et al.
Journal of chromatography. B, Biomedical sciences and applications, 755(1-2), 73-84 (2001-06-08)
Urinary metabolic ratios of caffeine are used in humans to assess the enzymatic activities of cytochrome P450 isoenzyme 1A2 (CYP1A2), xanthine oxidase (XO) and for phenotyping individuals for the bimodal N-acetyltransferase 2 (NAT2), all of them involved in the activation
Irene Zelner et al.
Drug metabolism reviews, 45(3), 277-299 (2013-05-30)
The ability to undergo non-oxidative metabolism from ethanol to fatty acid ethyl esters (FAEEs) varies greatly among tissues and organs. To gain a greater understanding of non-oxidative ethanol metabolism to FAEE, we aimed to collect all published data on FAEE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service