Skip to Content
Merck

D-110

N-Desmethyl-cis-tramadol-D3 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H20D3NO2 · HCl
CAS Number:
Molecular Weight:
288.83
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O[C@@]1(CCCC[C@@H]1CNC([2H])([2H])[2H])C2=CC(OC)=CC=C2.[H]Cl

InChI

1S/C15H23NO2.ClH/c1-16-11-13-6-3-4-9-15(13,17)12-7-5-8-14(10-12)18-2;/h5,7-8,10,13,16-17H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1/i1D3;

InChI key

NOZLWRHUQJHIRG-UOYBXXRGSA-N

General description

N-Desmethyl-cis-tramadol is a primary urinary and plasma metabolite of the synthetic analgesic tramadol. Tramadol, sold under trade names Ultram, Conzip, and Ryzolt, relieves moderate to severe pain. This stable-labeled internal standard is suitable for the quantitation of N-Desmethyl-cis-tramadol in biological samples using GC/MS or LC/MS with numerous testing applications including forensic or clinical toxicology analysis, urine drug testing, or isotope dilution methods.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library