Skip to Content
Merck
All Photos(1)

Key Documents

D-011

Supelco

1,3-Dinitroglycerin solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2O7
CAS Number:
Molecular Weight:
182.09
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

OC(CO[N+]([O-])=O)CO[N+]([O-])=O

InChI

1S/C3H6N2O7/c6-3(1-11-4(7)8)2-12-5(9)10/h3,6H,1-2H2

InChI key

ASIGVDLTBLZXNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Dinitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution® is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.

Application

  • Population pharmacokinetics of nitroglycerin and of its two metabolites after a single 24-hour application of a nitroglycerin transdermal matrix delivery system: This study presents a comprehensive analysis on the population pharmacokinetics of nitroglycerin, focusing on its behavior and the behavior of its metabolites following the use of a transdermal delivery system. The findings contribute to understanding the systemic availability and action of nitroglycerin from such systems, which could be relevant for 1,3-Dinitroglycerin given its structural and functional similarities (Auclair et al., 1998).
  • Novel pharmacokinetic modelling of transdermal nitroglycerin: This research explores advanced pharmacokinetic models that describe the absorption and action of nitroglycerin when administered through transdermal patches. Such models are crucial for developing effective transdermal delivery systems for similar compounds like 1,3-Dinitroglycerin (Auclair et al., 1998).
  • Bioequivalence Comparison of Two Drug-in-Adhesive Transdermal Nitroglycerin Patches: This study compares the bioequivalence of two transdermal patches delivering nitroglycerin, highlighting the critical factors in the design and performance of transdermal systems that could similarly apply to 1,3-Dinitroglycerin (Harrison et al., 1996).
  • Percutaneous absorption of 1,3-dinitroglycerin and a trial of pharmacokinetic analysis: Directly relevant to 1,3-Dinitroglycerin, this study examines its percutaneous absorption characteristics, offering insights into its potential applications in transdermal delivery systems (Ogiso et al., 1990).
  • Analysis of solutions containing glutathione and inorganic nitrite: application to nitroglycerin metabolism studies: While primarily focused on nitroglycerin, this analysis could provide secondary insights into the metabolic pathways and interactions of 1,3-Dinitroglycerin with biomolecules like glutathione, which is crucial for understanding its pharmacological effects (Curry et al., 1987).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Haefeli et al.
Clinical pharmacology and therapeutics, 52(6), 590-596 (1992-12-01)
Independent of the route of nitroglycerin administration, substantial amounts of 1,2-glyceryl dinitrate (1,2-GDN) and 1,3-glyceryl dinitrate (1,3-GDN) metabolites accumulate in humans. Thus far their pharmacologic activity in comparison to nitroglycerin in humans is unknown. To compare the venodilatory potency of
M V Wenzl et al.
British journal of pharmacology, 156(8), 1248-1255 (2009-03-04)
Vascular tolerance to nitroglycerin (GTN) may be caused by impaired GTN bioactivation due to inactivation of mitochondrial aldehyde dehydrogenase (ALDH2). As relaxation to GTN is reduced but still sensitive to ALDH2 inhibitors in ascorbate deficiency, we compared the contribution of
N Higo et al.
Pharmaceutical research, 9(2), 187-190 (1992-02-01)
The metabolism of nitroglycerin (GTN) to 1,2- and 1,3-glyceryl dinitrate (GDN) by hairless mouse skin in vitro has been measured. In the first set of experiments, GTN was incubated with the 9000g supernatant of fresh, homogenized tissue in the presence
F W Lee et al.
Journal of pharmacokinetics and biopharmaceutics, 21(5), 533-550 (1993-10-01)
Intravenous infusions of nitroglycerin (GTN), 1,2-glyceryl dinitrate (1,2-GDN), and 1,3-glyceryl dinitrate (1,3-GDN) were given to four conscious dogs at 10 micrograms/min, 30 micrograms/min, 50 micrograms/min, and 70 micrograms/min of GTN and 20 micrograms/min and 100 micrograms/min of GDNs. The steady
Magnus Petersson et al.
Clinical physiology and functional imaging, 28(4), 229-234 (2008-04-04)
Patients with chronic heart failure (CHF) often require higher doses of nitroglycerin (glyceryl trinitrate, GTN) than patients with normal cardiac function to achieve a given haemodynamic goal. Two pathways leading to biotransformation of GTN have been characterized; a high-affinity pathway

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service