Skip to Content
Merck

C-207

Colchicine solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C22H25NO6
CAS Number:
Molecular Weight:
399.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

N([C@H]1CCc2c(c(c(c(c2)OC)OC)OC)c3c1c[c](c(cc3)OC)=O)C(=O)C

InChI

1S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-18(26-2)17(25)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1

InChI key

IAKHMKGGTNLKSZ-INIZCTEOSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

42.8 °F - closed cup

flash_point_c

6.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library