Skip to Content
Merck

C-140

Cannabidivarin solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

CBDV solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H26O2
CAS Number:
Molecular Weight:
286.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

cannabis testing

format

single component solution

storage temp.

−70°C

SMILES string

Oc1c(c(cc(c1)CCC)O)C2C(CCC(=C2)C)C(=C)C

InChI

1S/C19H26O2/c1-5-6-14-10-17(20)19(18(21)11-14)16-9-13(4)7-8-15(16)12(2)3/h9-11,15-16,20-21H,2,5-8H2,1,3-4H3

InChI key

REOZWEGFPHTFEI-UHFFFAOYSA-N

General description

This certified Snap-N-Spike® solution is suitable for cannabidivarin testing methods by GC/MS, LC/MS or HPLC for applications in clinical toxicology, testing of Cannabis potency or impurity profiling, pharmaceutical research, forensic analysis, and urine drug testing. Cerilliant® solution Certified Reference Materials (CRMs) of cannabinoids are supplied in a convenient, quantitative, US DEA-exempt solution format and with TK#s for Canadian customers.

Cannabidivarin (CBDV) is a non-psychoactive phytocannabinoid that belongs to the class of C21 terpenophenolic compounds. It is found in Cannabis that reportedly has therapeutic efficacy in the treatment of pain, mood disorders, and inflammatory diseases.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Determination of eight cannabinoids in eight hemp samples and 12 forensic cannabis samples using reversed phase-high-pressure liquid chromatography (RP-HPLC) in combination with UV detector
  • Analysis of 13 oil samples obtained from hemp for seven major cannabinoids using high-performance liquid chromatography (HPLC) combined with UV detection
  • Development of a rapid liquid chromatographic (LC) method for the separation and identification of cannabidiol, cannabidivarin, Δ9-tetrahydrocannabivarin, and cannabigerol in mouse tissue samples in combination with tandem mass spectrometry (MS/MS)
  • Detection and quantification of cannabidiol, tetrahydrocannabinol, cannabinol, tetrahydrocannabivarin, cannabidivarin, and cannabigerol by liquid chromatography coupled with mass spectrometry in positive electrospray ionization (ESI) mode from hemp plants
  • Development and validation of an LC method combined with a UV detector for impurity analysis of cannabidivarin and cannabidibutol in cannabidiol obtained from hemp extracts

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

WGK

WGK 2

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library