Skip to Content
Merck
All Photos(1)

Documents

C-035

Supelco

(±)-Cotinine-d3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

(+/-)-Cotinine-d3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9D3N2O
CAS Number:
Molecular Weight:
179.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/i1D3

InChI key

UIKROCXWUNQSPJ-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cotinine is an alkaloid found in tobacco as well as a major urinary metabolite of nicotine. This stable-labeled internal standard is suitable for quantitation of cotinine levels in urine, blood, and saliva for GC/MS or LC-MS/MS applications in nicotine testing or clinical and diagnostic testing of nicotine biomarkers.

Application


  • (±)-Cotinine-d₃ solution in nicotine metabolism research: This stable isotope-labeled compound is utilized in the HPLC-MS analysis of nicotine and cotinine across various biological matrices, highlighting its critical role in advancing our understanding of nicotine metabolism and exposure in biomedical studies (Jin et al., 2022).

  • Biomarker analysis in passive smoking: Cotinine-d₃ serves as an internal standard in the precise quantification of urinary cotinine, aiding in the assessment of second-hand smoke exposure, particularly in pediatric health studies (Wang et al., 2014).

  • Enhanced analytical techniques for smoking cessation: Employed in a sophisticated UPLC-MS/MS setup, cotinine-d₃ is crucial for quantifying smoking-related biomarkers in clinical trials aimed at evaluating smoking cessation interventions, ensuring accurate and sensitive biomarker tracking (Dobrinas et al., 2011).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service