Skip to Content
Merck

B-005

Butalbital-D5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Butalbital-D5 solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11D5H11N2O3
CAS Number:
Molecular Weight:
229.29
MDL number:
UNSPSC Code:
41116107
EC Number:
200-659-6
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

N1C(=O)NC(=O)C(C1=O)(CCC(C)C)C(C(=C([2H])[2H])[2H])([2H])[2H]

InChI

1S/C12H18N2O3/c1-4-6-12(7-5-8(2)3)9(15)13-11(17)14-10(12)16/h4,8H,1,5-7H2,2-3H3,(H2,13,14,15,16,17)/i1D2,4D,6D2

InChI key

OFGPEYHYGSLQGU-LDEUCTAFSA-N

General description

A certified analytical reference solution and internal standard applicable for use in butalbital testing methods by GC/MS or LC/MS for applications in clinical toxicology, urine drug testing or forensic analysis. Butalbital is a barbiturate often combined with other medications such as acetaminophen or aspirin for the treatment of pain or codeine for headaches. Trade names of several butalbital combinations include Axocet®, Bucet, and Bupap® with acetaminophen and Axotal with aspirin.

Legal Information

Axocet is a registered trademark of Fougera Pharmaceuticals Inc.
Bupap is a registered trademark of ECR Pharmaceuticals Co., Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Butalbital-D5 solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
B-005-1ML04061833419021
FR180380018-104061839124103
FR180380018-1004061839124110
FR180380018-100004061839124134
FR180380018-10004061839124127