Skip to Content
Merck

Skip To

A-197

Ashwagandha Dietary Ingredients Mix solution

20-100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

composition

Withaferin A, 100 μg/mL , Withanolide A, 100 μg/mL , Withanolide B, 20 μg/mL , Withanoside V, 100 μg/mL , Withanoside IV, 100 μg/mL

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

20-100 μg/mL in methanol

format

multi-component solution

storage temp.

−70°C

General description

This phytochemical certified reference materials mix is produced at the Millipore Sigma Cerilliant site in Round Rock Texas under ISO/IEC 17025 and ISO 17034 accreditation and offers a cost efficient and precise tool for calibration and verification of multiple key components for Withania, a plant frequently used in traditonal indian medicine (Ayurveda). A comprehensive certificate including traceable values and expanded uncertainties for all the analytes is available.

genus withania

Discover the entire range of phytochemical certified reference material solutions from Cerilliant

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A-205CRM46304B-041
grade

certified reference material

grade

certified reference material

grade

certified reference material, TraceCERT®

grade

certified reference material

format

multi-component solution

format

single component solution

format

multi-component solution

format

multi-component solution

storage temp.

−70°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

composition

Withaferin A, 100 μg/mL , Withanolide B, 20 μg/mL , Withanoside IV, 100 μg/mL , Withanolide A, 100 μg/mL , Withanoside V, 100 μg/mL

composition

-

composition

-

composition

Amobarbital , Butalbital , Pentobarbital , Phenobarbital , Secobarbital

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

-

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters