Skip to Content
Merck

Skip To

A-076

Multicomponent Alcohol Mix-100

100 μg/mL each component in H2O, ampule of 1.2 mL, certified reference material, Cerilliant®

Synonym(s):

Multi-component Alcohol Mix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

Acetone , Ethanol , Isopropanol , Methanol, ACS grade

packaging

ampule of 1.2 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL each component in H2O

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

multi-component solution

storage temp.

2-8°C

InChI

1S/C15H13N3O/c16-11-6-7-13-12(8-11)15(17-9-14(19)18-13)10-4-2-1-3-5-10/h1-8H,9,16H2,(H,18,19)/i1D,2D,3D,4D,5D

InChI key

OYOUQHVDCKOOAL-RALIUCGRSA-N

General description

A multicomponent calibration kit containing acetone, ethanol, isopropanol, and methanol at a concentration of 100 μg/mL. This kit is suitable for calibration of blood alcohol testing methods using GC Headspace.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

12 - Non Combustible Liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters