Skip to Content
Merck

A-005

(±)-Amphetamine-d5 (deuterium label on side chain) solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9D5H8N
CAS Number:
Molecular Weight:
140.24
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

[2H]C(N)(C([2H])([2H])[2H])C([2H])C1=CC=CC=C1

InChI

1S/C9H13N/c1-8(10)7-9-5-3-2-4-6-9/h2-6,8H,7,10H2,1H3/i1D3,7D,8D

InChI key

KWTSXDURSIMDCE-ZXZAAQOOSA-N

General description

Amphetamine, a stimulant of the phenethylamine class, is found in ADHD-treatment drugs such as Adderall®, Vyvanse®, and ProCentra®. Because of its stimulant effect, amphetamine is used recreationally as a performance-enhancer and illicit drug. This stable-labeled internal standard is suitable for quantitation of amphetamine levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or sports testing.

Legal Information

Adderall is a registered trademark of Richwood Pharmaceutical Company, Inc.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
ProCentra is a registered trademark of Outlook Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyvanse is a registered trademark of Shire, LLC


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library