Skip to Content
Merck
All Photos(1)

Key Documents

LM1000

Avanti

12:0-13:0 PC

Avanti Research - A Croda Brand LM1000, methanol solution

Synonym(s):

1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H66NO8P
CAS Number:
Molecular Weight:
635.85
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1000-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1000

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O)=O

InChI key

FCTBVSCBBWKZML-WJOKGBTCSA-N

General description

Phospholipids are major structural biomolecules found in biological membranes. Phosphocholine (PC) is an intermediate of phosphatidylcholine synthesis catalyzed by choline kinase.

Application

12:0-13:0 PC or 1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phosphocholine has been used:
  • as an internal standard in liquid chromatography/mass spectrometry (LC/MS) analysis
  • as a lipid standard for phospholipid quantification by MS
  • as an internal standard for the retention time recalibration during LC-MS data processing
  • as an MS grade lipid standard for lipid extraction from macrophages

Biochem/physiol Actions

Phospholipids participate in protein mediated intracellular and extracellular signaling processes.

Packaging

2 mL Amber Glass Sealed Ampule (LM1000-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akifumi Shiomi et al.
Bioscience, biotechnology, and biochemistry, 84(3), 583-593 (2019-11-26)
Fish cell lines are widely used for the studies of developmental biology, virology, biology of aging, and nutrition physiology. However, little is known about their physicochemical properties. Here, we report the phospholipid compositions and mechanical properties of cell membranes derived
Dinesh Kumar Barupal et al.
Scientific data, 5, 180263-180263 (2018-11-21)
Alzheimer's disease (AD) is a major public health priority with a large socioeconomic burden and complex etiology. The Alzheimer Disease Metabolomics Consortium (ADMC) and the Alzheimer Disease Neuroimaging Initiative (ADNI) aim to gain new biological insights in the disease etiology.
Shayne A Snider et al.
The Journal of biological chemistry, 293(29), 11600-11611 (2018-06-09)
Choline is an essential nutrient that is required for synthesis of the main eukaryote phospholipid, phosphatidylcholine. Macrophages are innate immune cells that survey and respond to danger and damage signals. Although it is well-known that energy metabolism can dictate macrophage
Rachel Fickes et al.
Rapid communications in mass spectrometry : RCM, 30(24), 2601-2606 (2016-10-01)
Structural analogs of the bioactive lipid 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoglycerol were synthesized with a xylitol polar head group and both diacyl and diether radyl groups. Mass spectral characterization of xylitol phospholipids (PX) was carried out using collisional activation and high-resolution mass measurements of
Bingpeng Yan et al.
International journal of molecular sciences, 20(23) (2019-11-30)
Enterovirus A71 (EV-A71) and coxsackievirus A16 (CV-A16) are the most common causes of hand, foot, and mouth disease. Severe EV-A71 and CV-A16 infections may be associated with life-threatening complications. However, the pathogenic mechanisms underlying these severe clinical and pathological features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service