Skip to Content
Merck
All Photos(1)

Key Documents

880130P

Avanti

18:1 PEG2000 PE

Avanti Research - A Croda Brand 880130P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 g (880130P-1g)
pkg of 1 × 25 mg (880130P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880130P

transition temp

transition temp 12.8 °C (melting transition in micelles)

solubility

soluble (soluble in ethanol at 5mg/mL, soluble in DMSO at 5mg/mL, soluble in Chloroform:Methanol (85:15) at 5mg/mL)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+]

InChI

1S/C133H260NO55P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-131(135)186-128-130(189-132(136)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)129-188-190(138,139)187-37-36-134-133(137)185-127-126-184-125-124-183-123-122-182-121-120-181-119-118-180-1

InChI key

QXTYPWABYAAAPJ-FCKVYVLXSA-N

General description

18:1 PEG2000 PE also known as 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (DOPE-mPEG2K), is a polyethylene glycol (PEG)-phospholipid conjugate.

Application

18:1 PEG2000 PE or 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] has been used as a component of liposome stock solution for depositing lipid bilayers on the flowcell surface. It may be used in lipid bilayer preparation.

Packaging

5 mL Clear Glass Sealed Ampule (880130P-25mg)
60 mL Amber Wide Mouth Screw Cap Glass Bottle (880130P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Michael M Soniat et al.
Methods in enzymology, 592, 259-281 (2017-07-03)
Homologous recombination (HR) is a universally conserved DNA double-strand break repair pathway. Single-molecule fluorescence imaging approaches have revealed new mechanistic insights into nearly all aspects of HR. These methods are especially suited for studying protein complexes because multicolor fluorescent imaging
Na Young Cheon et al.
Nucleic acids research, 47(16), 8337-8347 (2019-08-03)
DNA repair is critical for maintaining genomic integrity. Finding DNA lesions initiates the entire repair process. In human nucleotide excision repair (NER), XPC-RAD23B recognizes DNA lesions and recruits downstream factors. Although previous studies revealed the molecular features of damage identification
Luisina De Tullio et al.
Methods in enzymology, 600, 407-437 (2018-02-21)
Helicases are crucial participants in many types of DNA repair reactions, including homologous recombination. The properties of these enzymes can be assayed by traditional bulk biochemical analysis; however, these types of assays cannot directly access some types of information. In
Carol Cho et al.
Nature communications, 10(1), 5764-5764 (2019-12-19)
The fundamental unit of chromatin, the nucleosome, is an intricate structure that requires histone chaperones for assembly. ATAD2 AAA+ ATPases are a family of histone chaperones that regulate nucleosome density and chromatin dynamics. Here, we demonstrate that the fission yeast ATAD2
Saki Yamaura et al.
Clinica chimica acta; international journal of clinical chemistry, 481, 184-188 (2018-03-20)
Measurement of lipoprotein-associated phospholipase A2 (Lp-PLA2) can be used as an adjunct to traditional cardiovascular risk factors for identifying individuals at higher risk of cardiovascular events. This can be performed by quantification of the protein concentration using an ELISA platform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service