Skip to Content
Merck
All Photos(1)

Key Documents

880128C

Avanti

DSPE-PEG(2000) Amine

Avanti Research - A Croda Brand 880128C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (880128C-10mg)
pkg of 1 × 2.5 mL (880128C-25mg)
pkg of 1 × 20 mL (880128C-500mg)
pkg of 1 × 4 mL (880128C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880128C

concentration

10 mg/mL (880128C-10mg)
10 mg/mL (880128C-25mg)
25 mg/mL (880128C-100mg)
25 mg/mL (880128C-500mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\N)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.

General description

DSPE-PEG(2000) amine is a functionalized polyethylene glycol (PEG) conjugated lipid.

Application

DSPE-PEG(2000) Amine has been used:
  • to prepare amine-functionalized vesicles
  • to prepare small unilamellar vesicles
  • to prepare liposomes

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (880128C-500mg)
5 mL Clear Glass Sealed Ampule (880128C-100mg)
5 mL Clear Glass Sealed Ampule (880128C-10mg)
5 mL Clear Glass Sealed Ampule (880128C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga B Garbuzenko et al.
Theranostics, 9(26), 8362-8376 (2019-11-23)
Non-Small Cell Lung Carcinoma (NSCLC), is the most common type of lung cancer (more than 80% of all cases). Small molecule Tyrosine Kinase (TK) Inhibitors acting on the Epidermal Growth Factor Receptors (EGFRs) are standard therapies for patients with NSCLC
Landscape phage fusion protein-mediated targeting of nanomedicines enhances their prostate tumor cell association and cytotoxic efficiency
Jayanna PK, et al.
Nanomedicine: Nanotechnology, Biology, and Medicine, 6(4), 538-546 (2010)
Stabilization and immobilization of aquaporin reconstituted lipid vesicles for water purification
Sun G, et al.
Colloids and Surfaces. B, Biointerfaces, 102, 466-471 (2013)
Federico Perche et al.
Cancer biology & therapy, 13(12), 1205-1213 (2012-08-16)
To determine whether the spheroid culture can be used to evaluate drug efficacy, we have evaluated the toxicity of free or carrier-associated doxorubicin as a single drug or in combination with other antineoplastic agents using the spheroid cultures of drug-resistant
Impact of hapten presentation on antibody binding at lipid membrane interfaces
Jung H, et al.
Biophysical Journal, 94(8), 3094-3103 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service