Skip to Content
Merck
All Photos(1)

Key Documents

860827P

Avanti

18:1(2R-OH) Ceramide

Avanti Research - A Croda Brand 860827P, powder

Synonym(s):

N-(2′-(R)-hydroxyoleoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H69NO4
CAS Number:
Molecular Weight:
579.94
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860827P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860827P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCC/C=C\CCCCCCCC)=O)CO

General description

18:1(2R-OH) Ceramide is a unique ceramide containing 18:1C long chain base fatty acid (oleic acid)-with 2′-hydroxyl group in R configuration.

Biochem/physiol Actions

hFA-ceramides facilitate epidermal permeability barrier function.

Packaging

5 mL Amber Glass Screw Cap Vial (860827P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin Guo et al.
Journal of lipid research, 53(7), 1327-1335 (2012-04-21)
FA 2-hydroxylase (FA2H) is an NAD(P)H-dependent enzyme that initiates FA α oxidation and is also responsible for the biosynthesis of 2-hydroxy FA (2-OH FA)-containing sphingolipids in mammalian cells. The 2-OH FA is chiral due to the asymmetric carbon bearing the
Ana B Herrero et al.
Cancer research, 68(23), 9779-9787 (2008-12-03)
PM02734 is a novel synthetic antitumor drug that is currently in phase I clinical trials. To gain some insight into its mode of action, we used the yeast Saccharomyces cerevisiae as a model system. Treatment of S. cerevisiae with PM02734
Hiroko Hama
Biochimica et biophysica acta, 1801(4), 405-414 (2009-12-23)
2-Hydroxy fatty acids (hFA) are important components of a subset of mammalian sphingolipids. The presence of hFA in sphingolipids is best described in the nervous system, epidermis, and kidney. However, the literature also indicates that various hFA-sphingolipids are present in
Nathan L Alderson et al.
Journal of lipid research, 50(6), 1203-1208 (2009-01-28)
Sphingolipids are ubiquitous components of eukaryotic cells that regulate various cellular functions. In many cell types, a fraction of sphingolipids contain 2-hydroxy fatty acids, produced by fatty acid 2-hydroxylase (FA2H), as the N-acyl chain of ceramide [hydroxyl fatty acid (hFA)-sphingolipids].
Zdzislaw M Szulc et al.
Bioorganic & medicinal chemistry, 18(21), 7565-7579 (2010-09-21)
A straightforward method for the simultaneous preparation of (2S,3R,2'R)- and (2S,3R,2'S)-2'-hydroxy-ceramides (2'-OHCer) from (2S,3R)-sphingosine acetonide precursors and racemic mixtures of 2-hydroxy fatty acids (2-OHFAs) is described. The obtained 2'-OH-C4-, -C6-, -C12-, -C16-Cer and 2'-OH-C6-dhCer pairs of diastereoisomers were characterized thoroughly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service