Skip to Content
Merck
All Photos(1)

Key Documents

860824P

Avanti

24:0(2S-OH) Ceramide

Avanti Research - A Croda Brand 860824P, powder

Synonym(s):

N-(2′-(S)-hydroxylignoceroyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H83NO4
CAS Number:
Molecular Weight:
666.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860824P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860824P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@@H](O)CCCCCCCCCCCCCCCCCCCCCC)=O)CO

InChI

1S/C42H83NO4/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-25-27-29-31-33-35-37-41(46)42(47)43-39(38-44)40(45)36-34-32-30-28-26-24-16-14-12-10-8-6-4-2/h34,36,39-41,44-46H,3-33,35,37-38H2,1-2H3,(H,43,47)/b36-34+/t39-,40+,41-/m0/s1

InChI key

WAYLDHLWVYQNSQ-QJYCDPGTSA-N

General description

Ceramides containing 2-hydroxy fatty acids (hFA) are present in the surface epithelium of the skin. 24:0(2S-OH) Ceramide is a unique ceramide containing 24C long chain base fatty acid (lignoceric acid)-with 2′-hydroxyl group in S configuration. NAD(P)H-dependent enzyme, fatty acid 2-hydroxylase (FA2H) catalyzes the synthesis of hFA-ceramide. Ceramides containing hFA in epidermis help in permeability barrier function.

Packaging

5 mL Amber Glass Screw Cap Vial (860824P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel A Peñalva et al.
Biochimica et biophysica acta, 1838(3), 731-738 (2013-12-10)
Unique species of ceramide (Cer) with very-long-chain polyunsaturated fatty acid (VLCPUFA), mainly 28-32 carbon atoms, 4-5 double bonds, in nonhydroxy and 2-hydroxy forms (n-V Cer and h-V Cer, respectively), are generated in rat spermatozoa from the corresponding sphingomyelins during the
Stereospecificity of fatty acid 2-hydroxylase and differential functions of 2-hydroxy fatty acid enantiomers
Guo L, et al.
Journal of Lipid Research, 53(7), 1327-1335 (2012)
Normal fur development and sebum production depends on fatty acid 2-hydroxylase expression in sebaceous glands
Maier H, et al.
The Journal of Biological Chemistry, 286(29), 25922-25934 (2011)
Helena Maier et al.
The Journal of biological chemistry, 286(29), 25922-25934 (2011-06-02)
2-Hydroxylated fatty acid (HFA)-containing sphingolipids are abundant in mammalian skin and are believed to play a role in the formation of the epidermal barrier. Fatty acid 2-hydroxylase (FA2H), required for the synthesis of 2-hydroxylated sphingolipids in various organs, is highly
Lin Guo et al.
Journal of lipid research, 53(7), 1327-1335 (2012-04-21)
FA 2-hydroxylase (FA2H) is an NAD(P)H-dependent enzyme that initiates FA α oxidation and is also responsible for the biosynthesis of 2-hydroxy FA (2-OH FA)-containing sphingolipids in mammalian cells. The 2-OH FA is chiral due to the asymmetric carbon bearing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service