Skip to Content
Merck
All Photos(1)

Key Documents

860823P

Avanti

24:0(2R-OH) Ceramide

Avanti Research - A Croda Brand 860823P, powder

Synonym(s):

N-(2′-(R)-hydroxylignoceroyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H83NO4
CAS Number:
Molecular Weight:
666.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860823P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860823P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC/C=C/[C@@H](O)[C@@H](NC([C@H](O)CCCCCCCCCCCCCCCCCCCCCC)=O)CO

InChI

1S/C42H83NO4/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-25-27-29-31-33-35-37-41(46)42(47)43-39(38-44)40(45)36-34-32-30-28-26-24-16-14-12-10-8-6-4-2/h34,36,39-41,44-46H,3-33,35,37-38H2,1-2H3,(H,43,47)/b36-34+/t39-,40+,41+/m0/s1

InChI key

WAYLDHLWVYQNSQ-CFBMXOPXSA-N

General description

Ceramides containing 2-hydroxy fatty acids (hFA) are present in the surface epithelium of the skin. 24:0(2R-OH) Ceramide is a 2R hydroxylated lignoceric acid -containing sphingolipid. hFA-ceramides are synthesized by the enzyme fatty acid 2-hydroxylase (FA2H). hFA-ceramides aid in epidermal permeability barrier function.

Packaging

5 mL Amber Glass Screw Cap Vial (860823P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej Kováčik et al.
Langmuir : the ACS journal of surfaces and colloids, 33(11), 2890-2899 (2017-02-24)
Ceramides (Cer) based on 6-hydroxysphingosine are important components of the human skin barrier, the stratum corneum. Although diminished concentrations of 6-hydroxyCer have been detected in skin diseases such as atopic dermatitis, our knowledge on these unusual sphingolipids, which have only
Stereospecificity of fatty acid 2-hydroxylase and differential functions of 2-hydroxy fatty acid enantiomers
Guo L, et al.
Journal of Lipid Research, 53(7), 1327-1335 (2012)
Normal fur development and sebum production depends on fatty acid 2-hydroxylase expression in sebaceous glands
Maier H, et al.
The Journal of Biological Chemistry, 286(29), 25922-25934 (2011)
Daniel A Peñalva et al.
Biochimica et biophysica acta, 1838(3), 731-738 (2013-12-10)
Unique species of ceramide (Cer) with very-long-chain polyunsaturated fatty acid (VLCPUFA), mainly 28-32 carbon atoms, 4-5 double bonds, in nonhydroxy and 2-hydroxy forms (n-V Cer and h-V Cer, respectively), are generated in rat spermatozoa from the corresponding sphingomyelins during the
Fatty acid 2-Hydroxylation in mammalian sphingolipid biology
Hama H, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1801(4), 405-414 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service