Skip to Content
Merck
All Photos(1)

Key Documents

860562C

Avanti

18:1-12:0 Biotin PE

Avanti Research - A Croda Brand 860562C

Synonym(s):

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H83N4O10PS
CAS Number:
Molecular Weight:
903.20
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (860562C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860562C

concentration

1 mg/mL (860562C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C45H83N4O10PS/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-23-30-42(51)56-35-38(36-58-60(54,55)57-34-32-46)59-43(52)31-24-21-18-15-13-16-19-22-27-33-47-41(50)29-26-25-28-40-44-39(37-61-40)48-45(53)49-44/h9-10,38-40,44H,2-8,11-37,46H2,1H3,(H,47,50)(H,54,55)(H2,48,49,53)/b10-9-/t38-,39+,40+,44+/m1/s1

InChI key

HACDNRDNASOHSM-IBTAXAMTSA-N

Application

18:1-12:0 Biotin PE has been used:
  • as a ligand in phosphoethanolamine binding assay
  • in the generation of vesicle reconstituted with syntaxin-1A and Synaptosomal nerve-associated protein 25 (SNAP-25) vesicles
  • in the preparation of biotinylated liposomes

Biochem/physiol Actions

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phosphoethanolamine, biotin PE is majorly used in the generation of lipid vesicle in supported lipid bilayer. It is also used in generation of phospholipid vesicle.

Packaging

5 mL Clear Glass Sealed Ampule (860562C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TIRF imaging of Fc gamma receptor microclusters dynamics and signaling on macrophages during frustrated phagocytosis
Lin J, et al.
BMC Immunology, 17(1), 5-5 (2016)
Molecular mechanisms of synaptic vesicle priming by Munc13 and Munc18
Lai Y, et al.
Neuron, 95(3), 591-607 (2017)
Factor VIII C1 domain residues Lys 2092 and Phe 2093 contribute to membrane binding and cofactor activity
Meems H, et al.
Blood, 114(18), 3938-3946 (2009)
Toh B Gang et al.
The Journal of biological chemistry, 287(51), 43116-43125 (2012-11-10)
Human C-reactive protein (CRP) protects mice from lethal Streptococcus pneumoniae infection when injected into mice within the range of 6 h before to 2 h after the administration of pneumococci. Because CRP binds to phosphocholine-containing substances and subsequently activates the
Matthew Brecher et al.
Journal of virology, 86(1), 364-372 (2011-10-28)
Cellular entry of Ebola virus (EBOV), a deadly hemorrhagic fever virus, is mediated by the viral glycoprotein (GP). The receptor-binding subunit of GP must be cleaved (by endosomal cathepsins) in order for entry and infection to proceed. Cleavage appears to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service