Skip to Content
Merck
All Photos(1)

Key Documents

860506P

Avanti

C6 Ceramide (d18:1/6:0)

Avanti Research - A Croda Brand

Synonym(s):

N-hexanoyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H47NO3
CAS Number:
Molecular Weight:
397.63
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860506P-10mg)
pkg of 1 × 5 mg (860506P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C24H47NO3/c1-3-5-7-8-9-10-11-12-13-14-15-16-18-19-23(27)22(21-26)25-24(28)20-17-6-4-2/h18-19,22-23,26-27H,3-17,20-21H2,1-2H3,(H,25,28)/b19-18+

InChI key

NPRJSFWNFTXXQC-VHEBQXMUSA-N

General description

C6 Ceramide (d18:1/6:0) is a short chain and a cell permeable ceramide. It comprises amino alcoholic chain attached to fatty acyl chain via an amide linkage.

Application

C6 Ceramide (d18:1/6:0) has been used as a chemotherapeutic agent to test its anti-tumor effect in breast cancer cells and ovarian cancer cell lines. It has also been used to test chemo-sensitization effects in cancer cells.

Biochem/physiol Actions

C6 Ceramide (d18:1/6:0) is involved in inhibiting proliferation and inducing apoptosis. When used in combination with acid ceramidase inhibitor DM102, [(2R,3Z)-N-(1-hydroxyoctadec-3-en-2-yl)pivaloylamide], it favors cell death in human breast cancer cells lines. C6 Ceramide may serve as an adjunct in chemotherapy. It elicits anti-tumor effects via AKT (serine/threonine-specific protein kinase) dephosphorylation and α-tubulin acetylation.

Packaging

5 mL Amber Glass Screw Cap Vial (860506P-10mg)
5 mL Amber Glass Screw Cap Vial (860506P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C6-ceramide and targeted inhibition of acid ceramidase induce synergistic decreases in breast cancer cell growth
Flowers M, et al.
Breast Cancer Research and Treatment, 133(2), 447-458 (2012)
Short-chain C6 ceramide sensitizes AT406-induced anti-pancreatic cancer cell activity
Zhao X, et al.
Biochemical and Biophysical Research Communications, 479(2), 166-172 (2016)
Mechanisms involved in exogenous C2-and C6-ceramide-induced cancer cell toxicity
Fillet M, et al.
Biochemical Pharmacology, 65(10), 1633-1642 (2003)
C6 ceramide dramatically enhances docetaxel-induced growth inhibition and apoptosis in cultured breast cancer cells: a mechanism study
Yang L, et al.
Experimental Cell Research, 332(1), 47-59 (2015)
C Ji et al.
Oncogene, 29(50), 6557-6568 (2010-08-31)
New chemotherapy-enhancing strategies are needed for better cancer therapy. Previous studies suggest that exogenous cell-permeable C6 ceramide may be a useful adjunct to the anti-tumor effects of chemotherapeutic agents (such as Taxol) against multiple cancers. Here we demonstrate that exogenous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service